Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747593_m1
          See other HAT1 GE Assays ›
          Gene Symbol:
          Hat1
          Gene Name:
          histone acetyltransferase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 58022395 - 58070064 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747593_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009657.1 NP_001009657.1 4 - 5 342 110
          GenBank mRNA BC087663.1 - 4 - 5 342 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000801.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hat1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication-dependent nucleosome assembly
          DNA replication-independent nucleosome assembly
          response to nutrient
          histone H4 acetylation
          molecular_function
          H4 histone acetyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79631 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084841 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084841 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084841 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P149734 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1364 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836825 - -
          Affymetrix Rat Gene 1.0 ST Array 10836819 - -
          Affymetrix Rat Gene 1.0 ST Array 10836821 - -
          Affymetrix Rat Gene 1.0 ST Array 10836816 - -
          Affymetrix Rat Gene 1.0 ST Array 10836820 - -
          Affymetrix Rat Gene 1.0 ST Array 10836817 - -
          Affymetrix Rat Gene 1.0 ST Array 10836818 - -
          Affymetrix Rat Gene 1.0 ST Array 10836823 - -
          Affymetrix Rat Gene 1.0 ST Array 10836826 - -
          Affymetrix Rat Gene 1.0 ST Array 10836824 - -
          Affymetrix Rat Gene 1.0 ST Array 10836822 - -
          Affymetrix Rat Gene 1.1 ST Array 10836815 - -
          Rat Genome 230 2.0 Array 1393647_PM_at - -
          Rat Genome 230 2.0 Array 1393647_at - -
          Affymetrix HT Rat Focus Array 1393647_at - -
          Affymetrix HT Rat Focus Array 1393647_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300006071.rn.1 TC0300000801.rn.1 RTA TC
          Rat Expression Set 230 1393647_at - -
          Rat Expression Set 230 1393647_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836815 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21066822 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357840 - -
          RatRef-12 Expression BeadChip ILMN_53256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0