Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01748805_g1
          See other M6PR GE Assays ›
          Gene Symbol:
          M6pr LOC100909548
          Gene Name:
          mannose-6-phosphate receptor, cation dependent
          cation-dependent mannose-6-phosphate receptor-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.4: 155084755 - 155093896 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 88312689 - 88321836 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01748805_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: M6pr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007700.1 NP_001007701.1 6 - 7 865 60
          XM_008763247.1 XP_008761469.1 6 - 7 1917 60
          XM_008763248.1 XP_008761470.1 6 - 7 930 60
          GenBank mRNA BC079226.1 - 6 - 7 865 60
          Gene Symbol: LOC100909548
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003752100.1 XP_003752148.1 6 - 7 871 60
          XM_006257201.2 XP_006257263.1 6 - 7 1917 60
          XM_006257202.1 XP_006257264.1 6 - 7 1090 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene M6pr
          Click here to find siRNAs for Gene LOC100909548

          Gene Ontology Categories:

          Function(s) Process(es)

          mannose transmembrane transport
          mannose binding
          mannose transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067801 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120982 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067803 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P766292 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P260487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72191 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934773 - -
          Affymetrix Rat Gene 1.0 ST Array 10858492 - -
          Affymetrix Rat Gene 1.0 ST Array 10858493 - -
          Affymetrix Rat Gene 1.0 ST Array 10858491 - -
          Affymetrix Rat Gene 1.0 ST Array 10858490 - -
          Affymetrix Rat Gene 1.0 ST Array 10858488 - -
          Affymetrix Rat Gene 1.0 ST Array 10858494 - -
          Affymetrix Rat Gene 1.0 ST Array 10934774 - -
          Affymetrix Rat Gene 1.0 ST Array 10858489 - -
          Affymetrix Rat Gene 1.1 ST Array 10858487 - -
          Affymetrix Rat Gene 1.1 ST Array 10934772 - -
          Rat Genome 230 2.0 Array 1371495_at - -
          Rat Genome 230 2.0 Array 1371495_PM_at - -
          Affymetrix HT Rat Focus Array 1371495_at - -
          Affymetrix HT Rat Focus Array 1371495_PM_at - -
          Rat Expression Set 230 1371495_PM_at - -
          Rat Expression Set 230 1371495_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858487 - -
          Affymetrix Rat Gene 1.0 ST Array 10934772 - -
          Rat Genome U34 Set rc_AA943990_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21397924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365228 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352588 - -
          RatRef-12 Expression BeadChip ILMN_55491 - -
          RatRef-12 Expression BeadChip ILMN_69293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline