Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749232_g1
          See other SOX17 GE Assays ›
          Gene Symbol:
          Sox17
          Gene Name:
          SRY (sex determining region Y)-box 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 14890309 - 14895907 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749232_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107902.1 NP_001101372.1 3 - 4 610 63
          XM_006237809.2 XP_006237871.1 3 - 4 1610 63
          XM_006237812.2 XP_006237874.1 3 - 4 1429 63
          XM_008763533.1 XP_008761755.1 4 - 5 1279 63
          XM_008763534.1 XP_008761756.1 4 - 5 1282 63
          XM_008763535.1 XP_008761757.1 4 - 5 1029 63
          XM_008763536.1 XP_008761758.1 4 - 5 1026 63
          XM_008763537.1 XP_008761759.1 3 - 4 738 63
          XM_008763538.1 XP_008761760.1 2 - 3 764 63
          GenBank mRNA BC166978.1 - 3 - 4 1312 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000118.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          vasculogenesis
          endoderm formation
          inner cell mass cellular morphogenesis
          heart looping
          cardiogenic plate morphogenesis
          embryonic heart tube morphogenesis
          outflow tract morphogenesis
          negative regulation of Wnt signaling pathway involved in heart development
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          spermatogenesis
          gastrulation
          endoderm development
          endodermal cell fate determination
          biological_process
          positive regulation of gene expression
          rostrocaudal neural tube patterning
          signal transduction involved in regulation of gene expression
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          protein destabilization
          embryonic heart tube development
          cell migration involved in gastrulation
          regulation of mesodermal cell fate specification
          negative regulation of mesodermal cell fate specification
          mRNA transcription by RNA polymerase II
          regulation of cell differentiation
          positive regulation of cell differentiation
          positive regulation of protein catabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of embryonic development
          embryonic organ development
          embryonic foregut morphogenesis
          positive regulation of skeletal muscle tissue development
          stem cell differentiation
          stem cell fate specification
          protein stabilization
          canonical Wnt signaling pathway
          endocardium formation
          cardiac cell fate determination
          endocardial cell differentiation
          common bile duct development
          gall bladder development
          endodermal digestive tract morphogenesis
          renal system development
          regulation of stem cell proliferation
          negative regulation of canonical Wnt signaling pathway
          regulation of stem cell division
          regulation of cardiac cell fate specification
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          molecular_function
          DNA binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          beta-catenin binding
          transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P483712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P483712 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P483712 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P483712 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P499350 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P483712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867408 - -
          Affymetrix Rat Gene 1.0 ST Array 10867407 - -
          Affymetrix Rat Gene 1.1 ST Array 10867406 - -
          Rat Genome 230 2.0 Array 1392990_at - -
          Rat Genome 230 2.0 Array 1378935_PM_at - -
          Rat Genome 230 2.0 Array 1378935_at - -
          Rat Genome 230 2.0 Array 1392990_PM_at - -
          Rat Genome 230 2.0 Array 1390041_PM_at - -
          Rat Genome 230 2.0 Array 1390041_at - -
          Affymetrix HT Rat Focus Array 1390041_PM_at - -
          Affymetrix HT Rat Focus Array 1378935_PM_at - -
          Affymetrix HT Rat Focus Array 1392990_PM_at - -
          Affymetrix HT Rat Focus Array 1392990_at - -
          Affymetrix HT Rat Focus Array 1390041_at - -
          Affymetrix HT Rat Focus Array 1378935_at - -
          Affymetrix Rat Clariom S Assay PSR0500000797.rn.1 TC0500000118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000798.rn.1 TC0500000118.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500000796.rn.1 TC0500000118.rn.1 RTA TC
          Rat Expression Set 230 1390041_PM_at - -
          Rat Expression Set 230 1390041_at - -
          Rat Expression Set 230 1378935_at - -
          Rat Expression Set 230 1378935_PM_at - -
          Rat Expression Set 230 1392990_at - -
          Rat Expression Set 230 1392990_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867406 - -
          Rat Genome U34 Set rc_AA799800_at - -
          Rat Genome U34 Set rc_AI031004_at - -
          Rat Genome U34 Set rc_AI235684_at - -
          Rat Genome U34 Set rc_AI172568_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21128741 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0