Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749332_g1
          See other RPL11 GE Assays ›
          Gene Symbol:
          Rpl11
          Gene Name:
          ribosomal protein L11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 154390809 - 154394407 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749332_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025739.1 NP_001020910.1 1 - 2 486 69
          XM_006239224.2 XP_006239286.1 2 - 3 229 69
          GenBank mRNA BC097372.1 - 1 - 2 486 69
          FQ217185.1 - 2 - 3 166 69
          FQ218104.1 - 2 - 3 171 69
          FQ221293.1 - 2 - 3 170 69
          FQ221469.1 - 2 - 3 170 69
          FQ222336.1 - 2 - 3 170 69
          FQ222952.1 - 2 - 3 166 69
          X62146.1 - 2 - 3 167 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003853.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl11

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          translation
          protein targeting
          protein localization to nucleus
          ribosomal large subunit biogenesis
          RNA binding
          structural constituent of ribosome
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P070065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735132 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070065 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070065 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P070065 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205902 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700490856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70960 - -
          Agilent Rat cDNA Microarray (G4105A) 700490856 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880679 - -
          Affymetrix Rat Gene 1.0 ST Array 10927124 - -
          Affymetrix Rat Gene 1.0 ST Array 10880681 - -
          Affymetrix Rat Gene 1.0 ST Array 10880680 - -
          Affymetrix Rat Gene 1.0 ST Array 10880682 - -
          Affymetrix Rat Gene 1.1 ST Array 10880678 - -
          Affymetrix Rat Gene 1.1 ST Array 10927122 - -
          Rat Genome 230 2.0 Array 1371330_PM_at - -
          Rat Genome 230 2.0 Array 1371330_at - -
          Affymetrix HT Rat Focus Array 1371330_PM_at - -
          Affymetrix HT Rat Focus Array 1371330_at - -
          Affymetrix Rat Clariom S Assay PSR0500028998.rn.1 TC0500003853.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029001.rn.1 TC0500003853.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029003.rn.1 TC0500003853.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029000.rn.1 TC0500003853.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029002.rn.1 TC0500003853.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028999.rn.1 TC0500003853.rn.1 RTA TC
          Rat Expression Set 230 1371330_at - -
          Rat Expression Set 230 1371330_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880678 - -
          Rat Genome U34 Set X62146cds_at - -
          Rat Genome U34 Set X62146cds_g_at - -
          Rat Genome U34 Set rc_AI233163_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21742029 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0