Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750242_m1
          See other MYBL2 GE Assays ›
          Gene Symbol:
          Mybl2
          Gene Name:
          myeloblastosis oncogene-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 159421637 - 159450087 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mybl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106536.1 NP_001100006.1 3 - 4 379 57
          XM_006235525.2 XP_006235587.1 3 - 4 413 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002250.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription by RNA polymerase II
          mitotic spindle assembly
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P653701 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13416 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P653701 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003124 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P827359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841979 - -
          Affymetrix Rat Gene 1.0 ST Array 10841980 - -
          Affymetrix Rat Gene 1.0 ST Array 10841976 - -
          Affymetrix Rat Gene 1.0 ST Array 10841982 - -
          Affymetrix Rat Gene 1.0 ST Array 10841985 - -
          Affymetrix Rat Gene 1.0 ST Array 10841987 - -
          Affymetrix Rat Gene 1.0 ST Array 10841975 - -
          Affymetrix Rat Gene 1.0 ST Array 10841983 - -
          Affymetrix Rat Gene 1.0 ST Array 10841981 - -
          Affymetrix Rat Gene 1.0 ST Array 10841984 - -
          Affymetrix Rat Gene 1.0 ST Array 10841978 - -
          Affymetrix Rat Gene 1.0 ST Array 10841977 - -
          Affymetrix Rat Gene 1.0 ST Array 10841986 - -
          Affymetrix Rat Gene 1.1 ST Array 10841974 - -
          Rat Genome 230 2.0 Array 1389457_at - -
          Rat Genome 230 2.0 Array 1377962_at - -
          Rat Genome 230 2.0 Array 1389457_PM_at - -
          Rat Genome 230 2.0 Array 1377962_PM_at - -
          Affymetrix HT Rat Focus Array 1389457_PM_at - -
          Affymetrix HT Rat Focus Array 1389457_at - -
          Affymetrix Rat Clariom S Assay PSR0300016772.rn.1 TC0300002250.rn.1 RTA TC
          Rat Expression Set 230 1389457_PM_at - -
          Rat Expression Set 230 1389457_at - -
          Rat Expression Set 230 1377962_PM_at - -
          Rat Expression Set 230 1377962_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841974 - -
          Rat Genome U34 Set rc_AA955126_at - -
          Rat Genome U34 Set rc_AI044979_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20760558 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline