Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750705_m1
          See other GJC1 GE Assays ›
          Gene Symbol:
          Gjc1
          Gene Name:
          gap junction protein, gamma 1
          Gene Aliases:
          Cx45, Gja7
          Chromosome Location:
          Chr.10: 90781674 - 90784359 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750705_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gjc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085381.1 NP_001078850.1 389 63
          GenBank mRNA BC166405.1 - 274 63

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC166405.1, NM_001085381.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          cell-cell signaling
          chemical synaptic transmission
          heart development
          visual perception
          ion transmembrane transport
          cell development
          cardiac muscle tissue development
          ion channel activity
          gap junction channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P504794 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504794 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P504794 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P504794 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173706 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P504794 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1384139_PM_at - -
          Rat Genome 230 2.0 Array 1384139_at - -
          Affymetrix Rat Clariom S Assay PSR1000035067.rn.1 TC1000003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035073.rn.1 TC1000003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035068.rn.1 TC1000003885.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035066.rn.1 TC1000003885.rn.1 RTA TC
          Rat Expression Set 230 1384139_at - -
          Rat Expression Set 230 1384139_PM_at - -
          Rat Genome U34 Set rc_AA956115_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21885050 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline