Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751636_m1
          See other SHMT1 GE Assays ›
          Gene Symbol:
          Shmt1
          Gene Name:
          serine hydroxymethyltransferase 1 (soluble)
          Gene Aliases:
          LRRGT00032, Shmt, mShmt
          Chromosome Location:
          Chr.10: 47030812 - 47059216 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751636_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047842.1 NP_001041307.1 13 - 14 1760 98
          XM_006246469.2 XP_006246531.1 10 - 11 1543 98
          GenBank mRNA AY383687.1 - 13 - 14 1760 98
          BC099219.1 - 6 - 7 774 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002942.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine metabolic process
          glycine biosynthetic process
          L-serine metabolic process
          L-serine catabolic process
          purine nucleobase biosynthetic process
          glycine biosynthetic process from serine
          tetrahydrofolate interconversion
          folic acid metabolic process
          protein tetramerization
          protein homotetramerization
          glycine hydroxymethyltransferase activity
          L-allo-threonine aldolase activity
          amino acid binding
          pyridoxal phosphate binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132367 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132367 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160813 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508239 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1055774 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14508 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10743321 - -
          Affymetrix Rat Gene 1.0 ST Array 10743331 - -
          Affymetrix Rat Gene 1.0 ST Array 10743327 - -
          Affymetrix Rat Gene 1.0 ST Array 10743332 - -
          Affymetrix Rat Gene 1.0 ST Array 10743325 - -
          Affymetrix Rat Gene 1.0 ST Array 10743322 - -
          Affymetrix Rat Gene 1.0 ST Array 10743320 - -
          Affymetrix Rat Gene 1.0 ST Array 10743333 - -
          Affymetrix Rat Gene 1.0 ST Array 10743319 - -
          Affymetrix Rat Gene 1.0 ST Array 10743330 - -
          Affymetrix Rat Gene 1.0 ST Array 10743328 - -
          Affymetrix Rat Gene 1.0 ST Array 10743324 - -
          Affymetrix Rat Gene 1.0 ST Array 10743329 - -
          Affymetrix Rat Gene 1.0 ST Array 10743323 - -
          Affymetrix Rat Gene 1.0 ST Array 10743326 - -
          Affymetrix Rat Gene 1.1 ST Array 10743318 - -
          Rat Genome 230 2.0 Array 1374748_at - -
          Rat Genome 230 2.0 Array 1373625_at - -
          Rat Genome 230 2.0 Array 1374748_PM_at - -
          Rat Genome 230 2.0 Array 1373625_PM_at - -
          Affymetrix HT Rat Focus Array 1373625_PM_at - -
          Affymetrix HT Rat Focus Array 1374748_at - -
          Affymetrix HT Rat Focus Array 1374748_PM_at - -
          Affymetrix HT Rat Focus Array 1373625_at - -
          Affymetrix Rat Clariom S Assay PSR1000025873.rn.1 TC1000002942.rn.1 RTA TC
          Rat Expression Set 230 1373625_PM_at - -
          Rat Expression Set 230 1373625_at - -
          Rat Expression Set 230 1374748_at - -
          Rat Expression Set 230 1374748_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10743318 - -
          Rat Genome U34 Set rc_AI011901_at - -
          Rat Genome U34 Set rc_AI012348_at - -
          Rat Genome U34 Set rc_AI011901_g_at - -
          Rat Genome U34 Set rc_AA819027_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20930758 - -
          ABI Rat Genome Survey Microarray 21771064 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0