Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751832_m1
          See other DUSP3 GE Assays ›
          Gene Symbol:
          Dusp3
          Gene Name:
          dual specificity phosphatase 3
          Gene Aliases:
          RGD1560049
          Chromosome Location:
          Chr.10: 89904578 - 89918308 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751832_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006247402.2 XP_006247464.1 3 - 4 515 51
          XM_006247403.2 XP_006247465.1 3 - 4 514 51
          GenBank mRNA BC169043.1 - 2 - 3 384 51

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003845.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          protein dephosphorylation
          peptidyl-tyrosine dephosphorylation
          negative regulation of MAPK cascade
          positive regulation of mitotic cell cycle
          negative regulation of JNK cascade
          negative regulation of T cell receptor signaling pathway
          negative regulation of T cell activation
          negative regulation of ERK1 and ERK2 cascade
          molecular_function
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein tyrosine/serine/threonine phosphatase activity
          MAP kinase phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10424 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P218284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P218284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P218284 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10424 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P218284 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P218284 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P729514 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747695 - -
          Affymetrix Rat Gene 1.0 ST Array 10747693 - -
          Affymetrix Rat Gene 1.0 ST Array 10747694 - -
          Affymetrix Rat Gene 1.1 ST Array 10747692 - -
          Rat Genome 230 2.0 Array 1377300_PM_at - -
          Rat Genome 230 2.0 Array 1377300_at - -
          Rat Genome 230 2.0 Array 1389050_at - -
          Rat Genome 230 2.0 Array 1389050_PM_at - -
          Affymetrix HT Rat Focus Array 1389050_at - -
          Affymetrix HT Rat Focus Array 1389050_PM_at - -
          Affymetrix HT Rat Focus Array 1377300_PM_at - -
          Affymetrix HT Rat Focus Array 1377300_at - -
          Affymetrix Rat Clariom S Assay PSR1000034608.rn.1 TC1000003845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034610.rn.1 TC1000003845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034607.rn.1 TC1000003845.rn.1 RTA TC
          Rat Expression Set 230 1389050_PM_at - -
          Rat Expression Set 230 1377300_PM_at - -
          Rat Expression Set 230 1377300_at - -
          Rat Expression Set 230 1389050_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747692 - -
          Rat Genome U34 Set rc_AI045719_at - -
          Rat Genome U34 Set rc_AI045130_at - -
          Rat Genome U34 Set rc_AI170797_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21400715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353786 - -
          RatRef-12 Expression BeadChip ILMN_64989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0