Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01751989_m1
          See other SMYD4 GE Assays ›
          Gene Symbol:
          Smyd4
          Gene Name:
          SET and MYND domain containing 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 62191669 - 62237959 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01751989_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smyd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105810.1 NP_001099280.1 5 - 6 1803 61
          XM_006246674.2 XP_006246736.1 5 - 6 1707 61
          XM_008767962.1 XP_008766184.1 4 - 5 1932 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001070.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P204207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P204207 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P204207 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P204207 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P204207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18823 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735886 - -
          Affymetrix Rat Gene 1.0 ST Array 10735884 - -
          Affymetrix Rat Gene 1.0 ST Array 10735881 - -
          Affymetrix Rat Gene 1.0 ST Array 10735877 - -
          Affymetrix Rat Gene 1.0 ST Array 10735880 - -
          Affymetrix Rat Gene 1.0 ST Array 10735878 - -
          Affymetrix Rat Gene 1.0 ST Array 10735879 - -
          Affymetrix Rat Gene 1.0 ST Array 10735883 - -
          Affymetrix Rat Gene 1.0 ST Array 10735885 - -
          Affymetrix Rat Gene 1.0 ST Array 10735882 - -
          Affymetrix Rat Gene 1.1 ST Array 10735876 - -
          Rat Genome 230 2.0 Array 1393116_PM_at - -
          Rat Genome 230 2.0 Array 1393116_at - -
          Affymetrix Rat Clariom S Assay PSR1000009674.rn.1 TC1000001070.rn.1 RTA TC
          Rat Expression Set 230 1393116_PM_at - -
          Rat Expression Set 230 1393116_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735876 - -
          Rat Genome U34 Set rc_AA964206_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22103905 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0