Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01752052_m1
          See other CD300LF GE Assays ›
          Gene Symbol:
          Cd300lf
          Gene Name:
          Cd300 molecule-like family member F
          Gene Aliases:
          CD300f, RGD1309733
          Chromosome Location:
          Chr.10: 103669781 - 103690577 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01752052_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd300lf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025111.1 NP_001020282.1 1 - 2 97 66
          XM_008768332.1 XP_008766554.1 2 - 3 953 66
          XM_008768333.1 XP_008766555.1 3 - 4 482 66
          XM_008768334.1 XP_008766556.1 2 - 3 372 66
          XM_008768337.1 XP_008766559.1 2 - 3 953 66
          XM_008768338.1 XP_008766560.1 2 - 3 951 66
          XM_008768339.1 XP_008766561.1 2 - 3 947 66
          XM_008768340.1 XP_008766562.1 2 - 3 950 66
          XM_008768341.1 XP_008766563.1 2 - 3 945 66
          XM_008768342.1 XP_008766564.1 2 - 3 942 66
          GenBank mRNA BC093593.1 - 1 - 2 97 66
          GU057984.1 - 1 - 2 65 66
          GU062903.1 - 1 - 2 62 66
          HQ263412.1 - 1 - 2 65 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004101.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd300lf

          Gene Ontology Categories:

          Function(s)

          osteoclast differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053675 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748871 - -
          Affymetrix Rat Gene 1.0 ST Array 10748880 - -
          Affymetrix Rat Gene 1.0 ST Array 10748874 - -
          Affymetrix Rat Gene 1.0 ST Array 10748881 - -
          Affymetrix Rat Gene 1.0 ST Array 10748873 - -
          Affymetrix Rat Gene 1.0 ST Array 10748876 - -
          Affymetrix Rat Gene 1.0 ST Array 10748879 - -
          Affymetrix Rat Gene 1.0 ST Array 10748878 - -
          Affymetrix Rat Gene 1.0 ST Array 10748872 - -
          Affymetrix Rat Gene 1.0 ST Array 10748875 - -
          Affymetrix Rat Gene 1.0 ST Array 10748877 - -
          Affymetrix Rat Gene 1.1 ST Array 10748870 - -
          Affymetrix Rat Clariom S Assay PSR1000037156.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037153.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037154.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037155.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10748870 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21096156 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0