Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01752474_m1
          See other RNF185 GE Assays ›
          Gene Symbol:
          Rnf185
          Gene Name:
          ring finger protein 185
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 83648400 - 83685273 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01752474_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf185
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024271.1 NP_001019442.1 6 - 7 635 94
          XM_006251333.1 XP_006251395.1 6 - 7 603 94
          XM_008770331.1 XP_008768553.1 4 - 5 447 94
          GenBank mRNA BC092655.1 - 6 - 7 635 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002126.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf185

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          protein ubiquitination
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P214687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P214687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P214687 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P214687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P214687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88181 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91241 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777952 - -
          Affymetrix Rat Gene 1.0 ST Array 10777957 - -
          Affymetrix Rat Gene 1.0 ST Array 10777954 - -
          Affymetrix Rat Gene 1.0 ST Array 10777953 - -
          Affymetrix Rat Gene 1.0 ST Array 10777951 - -
          Affymetrix Rat Gene 1.0 ST Array 10777955 - -
          Affymetrix Rat Gene 1.1 ST Array 10777950 - -
          Rat Genome 230 2.0 Array 1374701_PM_at - -
          Rat Genome 230 2.0 Array 1374701_at - -
          Affymetrix HT Rat Focus Array 1374701_at - -
          Affymetrix HT Rat Focus Array 1374701_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400015604.rn.1 TC1400002126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015606.rn.1 TC1400002126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015607.rn.1 TC1400002126.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015605.rn.1 TC1400002126.rn.1 RTA TC
          Rat Expression Set 230 1374701_at - -
          Rat Expression Set 230 1374701_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777950 - -
          Rat Genome U34 Set rc_AA943095_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366460 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0