Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753203_m1
          See other CRKL GE Assays ›
          Gene Symbol:
          Crkl LOC100911248
          Gene Name:
          v-crk avian sarcoma virus CT10 oncogene homolog-like
          crk-like protein-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.11: 87338606 - 87356644 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 87778338 - 87815051 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01753203_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crkl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008284.1 NP_001008285.1 1259 94
          GenBank mRNA BC085865.1 - 1259 94
          Gene Symbol: LOC100911248
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751095.3 XP_003751143.1 2 - 3 1291 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100002011.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crkl
          Click here to find siRNAs for Gene LOC100911248

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          heart development
          biological_process
          animal organ morphogenesis
          anterior/posterior pattern specification
          thymus development
          parathyroid gland development
          pattern specification process
          RNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150620 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45168 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156308 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156308 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156308 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166598 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97270 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48381 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755754 - -
          Affymetrix Rat Gene 1.0 ST Array 10755753 - -
          Affymetrix Rat Gene 1.0 ST Array 10755752 - -
          Affymetrix Rat Gene 1.1 ST Array 10755751 - -
          Rat Genome 230 2.0 Array 1393942_PM_at - -
          Rat Genome 230 2.0 Array 1395605_at - -
          Rat Genome 230 2.0 Array 1393942_at - -
          Rat Genome 230 2.0 Array 1395605_PM_at - -
          Affymetrix HT Rat Focus Array 1393942_at - -
          Affymetrix HT Rat Focus Array 1393942_PM_at - -
          Affymetrix HT Rat Focus Array 1395605_at - -
          Affymetrix HT Rat Focus Array 1395605_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100013273.rn.1 TC1100002011.rn.1 RTA TC
          Rat Expression Set 230 1393942_at - -
          Rat Expression Set 230 1395605_PM_at - -
          Rat Expression Set 230 1393942_PM_at - -
          Rat Expression Set 230 1395605_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755751 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21553001 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369034 - -
          RatRef-12 Expression BeadChip ILMN_57075 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0