Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753390_m1
          See other TNFRSF10B GE Assays ›
          Gene Symbol:
          Tnfrsf10b
          Gene Name:
          tumor necrosis factor receptor superfamily, member 10b
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 51433853 - 51464215 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753390_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf10b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108873.1 NP_001102343.1 2 - 3 359 59
          GenBank mRNA FJ515907.1 - 2 - 3 154 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000763.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          male gonad development
          extrinsic apoptotic signaling pathway via death domain receptors
          response to organic substance
          response to organic cyclic compound
          TRAIL-activated apoptotic signaling pathway
          cellular response to mechanical stimulus
          extrinsic apoptotic signaling pathway
          protease binding
          molecular_function
          transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095438 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095438 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095438 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781322 - -
          Affymetrix Rat Gene 1.0 ST Array 10781326 - -
          Affymetrix Rat Gene 1.0 ST Array 10781324 - -
          Affymetrix Rat Gene 1.0 ST Array 10781323 - -
          Affymetrix Rat Gene 1.0 ST Array 10781327 - -
          Affymetrix Rat Gene 1.0 ST Array 10781325 - -
          Affymetrix Rat Gene 1.1 ST Array 10781321 - -
          Affymetrix Rat Clariom S Assay PSR1500005282.rn.1 TC1500000763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005284.rn.1 TC1500000763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005285.rn.1 TC1500000763.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10781321 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22196802 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0