Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755810_m1
          See other ME2 GE Assays ›
          Gene Symbol:
          Me2
          Gene Name:
          malic enzyme 2, NAD(+)-dependent, mitochondrial
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 69738686 - 69784099 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755810_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Me2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107376.2 NP_001100846.2 13 - 14 1523 65
          XM_006254923.2 XP_006254985.1 13 - 14 4670 65
          GenBank mRNA CV796911.1 - 3 - 4 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001687.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          malate metabolic process
          malate dehydrogenase (decarboxylating) (NAD+) activity
          metal ion binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036049 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036049 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P752796 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372907 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805199 - -
          Affymetrix Rat Gene 1.0 ST Array 10805190 - -
          Affymetrix Rat Gene 1.0 ST Array 10805193 - -
          Affymetrix Rat Gene 1.0 ST Array 10805201 - -
          Affymetrix Rat Gene 1.0 ST Array 10805188 - -
          Affymetrix Rat Gene 1.0 ST Array 10805198 - -
          Affymetrix Rat Gene 1.0 ST Array 10805189 - -
          Affymetrix Rat Gene 1.0 ST Array 10805191 - -
          Affymetrix Rat Gene 1.0 ST Array 10805192 - -
          Affymetrix Rat Gene 1.0 ST Array 10805194 - -
          Affymetrix Rat Gene 1.0 ST Array 10805200 - -
          Affymetrix Rat Gene 1.0 ST Array 10805197 - -
          Affymetrix Rat Gene 1.0 ST Array 10805195 - -
          Affymetrix Rat Gene 1.0 ST Array 10805187 - -
          Affymetrix Rat Gene 1.0 ST Array 10805196 - -
          Affymetrix Rat Gene 1.1 ST Array 10805186 - -
          Rat Genome 230 2.0 Array 1393475_at - -
          Rat Genome 230 2.0 Array 1381403_PM_at - -
          Rat Genome 230 2.0 Array 1381403_at - -
          Rat Genome 230 2.0 Array 1393475_PM_at - -
          Affymetrix HT Rat Focus Array 1381403_at - -
          Affymetrix HT Rat Focus Array 1381403_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800011828.rn.1 TC1800001687.rn.1 RTA TC
          Rat Expression Set 230 1381403_PM_at - -
          Rat Expression Set 230 1381403_at - -
          Rat Expression Set 230 1393475_PM_at - -
          Rat Expression Set 230 1393475_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805186 - -
          Rat Genome U34 Set rc_AA944282_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21649006 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351957 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0