Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756123_m1
          See other NGLY1 GE Assays ›
          Gene Symbol:
          Ngly1
          Gene Name:
          N-glycanase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 10405453 - 10455973 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756123_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ngly1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014136.1 NP_001014158.1 10 - 11 1675 119
          GenBank mRNA BC083837.1 - 10 - 11 1675 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001333.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ngly1

          Gene Ontology Categories:

          Function(s) Process(es)

          glycoprotein catabolic process
          peptide-N4-(N-acetyl-beta-glucosaminyl)asparagine amidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P163266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P163266 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116809 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156741 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25602 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38930 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782477 - -
          Affymetrix Rat Gene 1.0 ST Array 10782480 - -
          Affymetrix Rat Gene 1.0 ST Array 10782484 - -
          Affymetrix Rat Gene 1.0 ST Array 10782473 - -
          Affymetrix Rat Gene 1.0 ST Array 10782475 - -
          Affymetrix Rat Gene 1.0 ST Array 10782472 - -
          Affymetrix Rat Gene 1.0 ST Array 10782479 - -
          Affymetrix Rat Gene 1.0 ST Array 10782483 - -
          Affymetrix Rat Gene 1.0 ST Array 10782478 - -
          Affymetrix Rat Gene 1.0 ST Array 10782476 - -
          Affymetrix Rat Gene 1.0 ST Array 10782474 - -
          Affymetrix Rat Gene 1.0 ST Array 10782481 - -
          Affymetrix Rat Gene 1.0 ST Array 10782482 - -
          Affymetrix Rat Gene 1.1 ST Array 10782471 - -
          Rat Genome 230 2.0 Array 1384205_at - -
          Rat Genome 230 2.0 Array 1384205_PM_at - -
          Affymetrix HT Rat Focus Array 1384205_at - -
          Affymetrix HT Rat Focus Array 1384205_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500008434.rn.1 TC1500001333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500008436.rn.1 TC1500001333.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500008433.rn.1 TC1500001333.rn.1 RTA TC
          Rat Expression Set 230 1384205_PM_at - -
          Rat Expression Set 230 1384205_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782471 - -
          Rat Genome U34 Set rc_AI010637_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349186 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0