Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756213_m1
          See other PTPN20B GE Assays ›
          Gene Symbol:
          Ptpn20b
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 20B
          Gene Aliases:
          Ptpn20
          Chromosome Location:
          Chr.16: 9907607 - 9973514 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756213_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptpn20b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080149.1 NP_001073618.1 9 - 10 1321 77
          XM_006252765.2 XP_006252827.1 10 - 11 1394 77
          XM_006252767.2 XP_006252829.1 10 - 11 1361 77
          XM_008771099.1 XP_008769321.1 10 - 11 1347 77
          GenBank mRNA BC129117.1 - 9 - 10 1321 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000131.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine dephosphorylation
          protein tyrosine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P137893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137893 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P137893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P137893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77562 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786897 - -
          Affymetrix Rat Gene 1.0 ST Array 10786894 - -
          Affymetrix Rat Gene 1.0 ST Array 10786902 - -
          Affymetrix Rat Gene 1.0 ST Array 10786895 - -
          Affymetrix Rat Gene 1.0 ST Array 10786896 - -
          Affymetrix Rat Gene 1.0 ST Array 10786903 - -
          Affymetrix Rat Gene 1.0 ST Array 10786901 - -
          Affymetrix Rat Gene 1.0 ST Array 10786900 - -
          Affymetrix Rat Gene 1.0 ST Array 10786898 - -
          Affymetrix Rat Gene 1.0 ST Array 10786899 - -
          Affymetrix Rat Gene 1.0 ST Array 10786904 - -
          Affymetrix Rat Gene 1.1 ST Array 10786893 - -
          Affymetrix Rat Clariom S Assay PSR1600001537.rn.1 TC1600000131.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001534.rn.1 TC1600000131.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001535.rn.1 TC1600000131.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001533.rn.1 TC1600000131.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001536.rn.1 TC1600000131.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10786893 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22088511 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361962 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0