Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756801_m1
          See other XKR5 GE Assays ›
          Gene Symbol:
          Xkr5
          Gene Name:
          XK, Kell blood group complex subunit-related family, member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 75744786 - 75762694 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756801_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xkr5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012228.1 NP_001012228.1 4 - 5 453 96
          GenBank mRNA AY534259.1 - 4 - 5 453 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000909.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xkr5

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P671422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137146 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671422 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671422 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P671422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81803 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789289 - -
          Affymetrix Rat Gene 1.0 ST Array 10789287 - -
          Affymetrix Rat Gene 1.0 ST Array 10789288 - -
          Affymetrix Rat Gene 1.0 ST Array 10789282 - -
          Affymetrix Rat Gene 1.0 ST Array 10789290 - -
          Affymetrix Rat Gene 1.1 ST Array 10789281 - -
          Rat Genome 230 2.0 Array 1376381_PM_at - -
          Rat Genome 230 2.0 Array 1376381_at - -
          Affymetrix HT Rat Focus Array 1376381_PM_at - -
          Affymetrix HT Rat Focus Array 1376381_at - -
          Affymetrix Rat Clariom S Assay PSR1600006989.rn.1 TC1600000909.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600006990.rn.1 TC1600000909.rn.1 RTA TC
          Rat Expression Set 230 1376381_PM_at - -
          Rat Expression Set 230 1376381_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789281 - -
          Rat Genome U34 Set rc_AI045409_s_at - -
          Rat Genome U34 Set rc_AI072208_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20880500 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0