Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757146_m1
          See other LOC501110 GE Assays ›
          Gene Symbol:
          LOC501110
          Gene Name:
          similar to Glutathione S-transferase A1 (GTH1) (HA subunit 1) (GST-epsilon) (GSTA1-1) (GST class-alpha)
          Gene Aliases:
          Gsta6
          Chromosome Location:
          Chr.9: 27475273 - 27533939 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757146_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC501110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024361.1 NP_001019532.1 1 - 2 113 71
          XM_006244656.2 XP_006244718.1 2 - 3 216 71
          XM_006244657.2 XP_006244719.1 2 - 3 183 71
          XM_006244658.2 XP_006244720.1 2 - 3 565 71
          GenBank mRNA BC079271.1 - 1 - 2 113 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001715.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC079271.1, NM_001024361.1, XM_006244656.2, XM_006244657.2, XM_006244658.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC501110

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          epithelial cell differentiation
          glutathione transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P623733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P623733 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P623733 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P623733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78425 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926968 - -
          Affymetrix Rat Gene 1.0 ST Array 10926969 - -
          Affymetrix Rat Gene 1.0 ST Array 10926972 - -
          Affymetrix Rat Gene 1.0 ST Array 10926974 - -
          Affymetrix Rat Gene 1.0 ST Array 10926970 - -
          Affymetrix Rat Gene 1.0 ST Array 10926971 - -
          Affymetrix Rat Gene 1.0 ST Array 10926973 - -
          Affymetrix Rat Gene 1.1 ST Array 10926967 - -
          Affymetrix Rat Clariom S Assay PSR0900013991.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013994.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013995.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013999.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013993.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013997.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013990.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013998.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013992.rn.1 TC0900001715.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10926967 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21029681 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357460 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0