Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01759635_m1
          See other DTX4 GE Assays ›
          Gene Symbol:
          Dtx4
          Gene Name:
          deltex 4, E3 ubiquitin ligase
          Gene Aliases:
          LRRGT00037
          Chromosome Location:
          Chr.1: 228730081 - 228759781 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01759635_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dtx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047855.1 NP_001041320.1 7 - 8 739 62
          XM_006231114.2 XP_006231176.1 8 - 9 2560 62
          GenBank mRNA AY383692.1 - 7 - 8 739 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007310.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P221437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99270 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P221437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P221437 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P221437 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P221434 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P221437 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51726 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729016 - -
          Affymetrix Rat Gene 1.0 ST Array 10729013 - -
          Affymetrix Rat Gene 1.0 ST Array 10729017 - -
          Affymetrix Rat Gene 1.0 ST Array 10729011 - -
          Affymetrix Rat Gene 1.0 ST Array 10729015 - -
          Affymetrix Rat Gene 1.0 ST Array 10729012 - -
          Affymetrix Rat Gene 1.0 ST Array 10729014 - -
          Affymetrix Rat Gene 1.1 ST Array 10729009 - -
          Rat Genome 230 2.0 Array 1398705_at - -
          Rat Genome 230 2.0 Array 1398705_PM_at - -
          Rat Genome 230 2.0 Array 1377982_PM_at - -
          Rat Genome 230 2.0 Array 1377982_at - -
          Affymetrix HT Rat Focus Array 1377982_at - -
          Affymetrix HT Rat Focus Array 1398705_at - -
          Affymetrix HT Rat Focus Array 1377982_PM_at - -
          Affymetrix HT Rat Focus Array 1398705_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100059185.rn.1 TC0100007310.rn.1 RTA TC
          Rat Expression Set 230 1398705_PM_at - -
          Rat Expression Set 230 1398705_at - -
          Rat Expression Set 230 1377982_at - -
          Rat Expression Set 230 1377982_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729009 - -
          Rat Genome U34 Set rc_AI177912_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21649505 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0