Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01760036_m1
          See other HHEX GE Assays ›
          Gene Symbol:
          Hhex
          Gene Name:
          hematopoietically expressed homeobox
          Gene Aliases:
          Hex
          Chromosome Location:
          Chr.1: 256101994 - 256107581 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01760036_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hhex
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024385.1 NP_077361.1 3 - 4 602 83
          GenBank mRNA BC088135.1 - 3 - 4 607 83
          D86383.1 - 3 - 4 602 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003551.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hhex

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          vasculogenesis
          in utero embryonic development
          liver development
          morphogenesis of an epithelium
          myeloid leukocyte differentiation
          regulation of transcription by RNA polymerase II
          mRNA export from nucleus
          signal transduction
          endoderm development
          cell population proliferation
          response to wounding
          animal organ morphogenesis
          anterior/posterior pattern specification
          negative regulation of angiogenesis
          poly(A)+ mRNA export from nucleus
          interkinetic nuclear migration
          hemopoiesis
          cell differentiation
          positive regulation of Wnt signaling pathway
          B cell differentiation
          thyroid gland development
          forebrain development
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          pancreas development
          protein localization to nucleus
          embryonic heart tube development
          multicellular organism growth
          regulation of cell population proliferation
          response to peptide hormone
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          embryonic organ development
          tissue morphogenesis
          forebrain morphogenesis
          primary lung bud formation
          common bile duct development
          gall bladder development
          hepatic duct development
          hepatoblast differentiation
          hepatocyte differentiation
          DNA conformation change
          primitive streak formation
          transcription cis-regulatory region binding
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription repressor activity, RNA polymerase II-specific
          chromatin binding
          DNA-binding transcription factor activity
          transcription factor binding
          eukaryotic initiation factor 4E binding
          DNA binding, bending
          protein homodimerization activity
          sequence-specific DNA binding
          HMG box domain binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162711 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12437 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12437 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12437 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700696715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90602 - -
          Agilent Rat cDNA Microarray (G4105A) 700696715 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715104 - -
          Affymetrix Rat Gene 1.0 ST Array 10715103 - -
          Affymetrix Rat Gene 1.0 ST Array 10715101 - -
          Affymetrix Rat Gene 1.0 ST Array 10715102 - -
          Affymetrix Rat Gene 1.1 ST Array 10715100 - -
          Rat Genome 230 2.0 Array 1387270_PM_at - -
          Rat Genome 230 2.0 Array 1387270_at - -
          Affymetrix HT Rat Focus Array 1387270_PM_at - -
          Affymetrix HT Rat Focus Array 1387270_at - -
          Affymetrix Rat Clariom S Assay PSR0100028821.rn.1 TC0100003551.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028820.rn.1 TC0100003551.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028822.rn.1 TC0100003551.rn.1 RTA TC
          Rat Expression Set 230 1387270_at - -
          Rat Expression Set 230 1387270_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715100 - -
          Rat Genome U34 Set rc_AA996940_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22295393 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353851 - -
          RatRef-12 Expression BeadChip ILMN_57808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0