Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01760991_m1
          See other REEP5 GE Assays ›
          Gene Symbol:
          Reep5
          Gene Name:
          receptor accessory protein 5
          Gene Aliases:
          Dp1
          Chromosome Location:
          Chr.18: 27128462 - 27159693 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01760991_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reep5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108888.2 NP_001102358.2 3 - 4 445 59
          GenBank mRNA BC167013.1 - 3 - 4 370 59
          FQ213789.1 - 3 - 4 377 59
          FQ235123.1 - 3 - 4 377 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001220.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P941342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34120 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P941342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P941342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P941342 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433771 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803731 - -
          Affymetrix Rat Gene 1.0 ST Array 10803733 - -
          Affymetrix Rat Gene 1.0 ST Array 10803729 - -
          Affymetrix Rat Gene 1.0 ST Array 10803730 - -
          Affymetrix Rat Gene 1.0 ST Array 10803732 - -
          Affymetrix Rat Gene 1.1 ST Array 10803728 - -
          Rat Genome 230 2.0 Array 1371481_at - -
          Rat Genome 230 2.0 Array 1371481_PM_at - -
          Rat Genome 230 2.0 Array 1371711_PM_at - -
          Rat Genome 230 2.0 Array 1371711_at - -
          Affymetrix HT Rat Focus Array 1371711_at - -
          Affymetrix HT Rat Focus Array 1371481_at - -
          Affymetrix HT Rat Focus Array 1371711_PM_at - -
          Affymetrix HT Rat Focus Array 1371481_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800008521.rn.1 TC1800001220.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008520.rn.1 TC1800001220.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008519.rn.1 TC1800001220.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008518.rn.1 TC1800001220.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800008517.rn.1 TC1800001220.rn.1 RTA TC
          Rat Expression Set 230 1371711_PM_at - -
          Rat Expression Set 230 1371481_PM_at - -
          Rat Expression Set 230 1371481_at - -
          Rat Expression Set 230 1371711_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803728 - -
          Rat Genome U34 Set rc_AA848813_g_at - -
          Rat Genome U34 Set rc_AA848813_at - -
          Rat Genome U34 Set rc_AI044771_at - -
          Rat Genome U34 Set rc_AI176346_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20765504 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352309 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0