Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761797_g1
          See other KRT83 GE Assays ›
          Gene Symbol:
          Krt83
          Gene Name:
          keratin 83, type II
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 143078996 - 143085833 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761797_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101675.1 NP_001095145.1 8 - 9 1299 113
          GenBank mRNA AB354637.1 - 8 - 9 1299 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003935.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          biological_process
          hair cycle
          molecular_function
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23722 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120818 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120818 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102986 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907363 - -
          Affymetrix Rat Gene 1.0 ST Array 10907358 - -
          Affymetrix Rat Gene 1.0 ST Array 10907359 - -
          Affymetrix Rat Gene 1.0 ST Array 10907360 - -
          Affymetrix Rat Gene 1.0 ST Array 10907362 - -
          Affymetrix Rat Gene 1.0 ST Array 10907361 - -
          Affymetrix Rat Gene 1.1 ST Array 10907357 - -
          Affymetrix Rat Clariom S Assay PSR0700031727.rn.1 TC0700003935.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031728.rn.1 TC0700003935.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10907357 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360970 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline