Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01762203_m1
          See other MGAT4D GE Assays ›
          Gene Symbol:
          Mgat4d
          Gene Name:
          MGAT4 family, member D
          Gene Aliases:
          RGD1310572
          Chromosome Location:
          Chr.19: 24494407 - 24532990 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01762203_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgat4d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025666.1 NP_001020837.1 10 - 11 1177 99
          XM_008772379.1 XP_008770601.1 10 - 11 1262 99
          XM_008772380.1 XP_008770602.1 10 - 11 1114 99
          GenBank mRNA BC097408.1 - 10 - 11 1177 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000291.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mgat4d

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          hexosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99487 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45248 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78292 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810331 - -
          Affymetrix Rat Gene 1.0 ST Array 10810326 - -
          Affymetrix Rat Gene 1.0 ST Array 10810327 - -
          Affymetrix Rat Gene 1.0 ST Array 10810324 - -
          Affymetrix Rat Gene 1.0 ST Array 10810328 - -
          Affymetrix Rat Gene 1.0 ST Array 10810332 - -
          Affymetrix Rat Gene 1.0 ST Array 10810329 - -
          Affymetrix Rat Gene 1.0 ST Array 10810323 - -
          Affymetrix Rat Gene 1.0 ST Array 10810325 - -
          Affymetrix Rat Gene 1.0 ST Array 10810330 - -
          Affymetrix Rat Gene 1.0 ST Array 10810333 - -
          Affymetrix Rat Gene 1.1 ST Array 10810322 - -
          Affymetrix Rat Clariom S Assay PSR1900002147.rn.1 TC1900000291.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10810322 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0