Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763041_m1
          See other ZFP282 GE Assays ›
          Gene Symbol:
          Zfp282
          Gene Name:
          zinc finger protein 282
          Gene Aliases:
          Znf282
          Chromosome Location:
          Chr.4: 77554269 - 77580714 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01763041_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp282
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106592.1 NP_001100062.1 7 - 8 1293 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000834.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P639174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724752 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P639174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P639174 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P639174 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55588 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855198 - -
          Affymetrix Rat Gene 1.0 ST Array 10855202 - -
          Affymetrix Rat Gene 1.0 ST Array 10855195 - -
          Affymetrix Rat Gene 1.0 ST Array 10855197 - -
          Affymetrix Rat Gene 1.0 ST Array 10855201 - -
          Affymetrix Rat Gene 1.0 ST Array 10855200 - -
          Affymetrix Rat Gene 1.0 ST Array 10855196 - -
          Affymetrix Rat Gene 1.0 ST Array 10855199 - -
          Affymetrix Rat Gene 1.1 ST Array 10855194 - -
          Rat Genome 230 2.0 Array 1389674_at - -
          Rat Genome 230 2.0 Array 1378996_at - -
          Rat Genome 230 2.0 Array 1378996_PM_at - -
          Rat Genome 230 2.0 Array 1389674_PM_at - -
          Affymetrix HT Rat Focus Array 1378996_at - -
          Affymetrix HT Rat Focus Array 1378996_PM_at - -
          Affymetrix HT Rat Focus Array 1389674_at - -
          Affymetrix HT Rat Focus Array 1389674_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400005837.rn.1 TC0400000834.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005834.rn.1 TC0400000834.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005836.rn.1 TC0400000834.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005835.rn.1 TC0400000834.rn.1 RTA TC
          Rat Expression Set 230 1389674_at - -
          Rat Expression Set 230 1389674_PM_at - -
          Rat Expression Set 230 1378996_PM_at - -
          Rat Expression Set 230 1378996_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855194 - -
          Rat Genome U34 Set rc_AA924770_at - -
          Rat Genome U34 Set rc_AA956035_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22000445 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363294 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0