Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763558_g1
          See other LOC103693563 GE Assays ›
          Gene Symbol:
          LOC103693563 Abhd10
          Gene Name:
          mycophenolic acid acyl-glucuronide esterase, mitochondrial
          abhydrolase domain containing 10
          Gene Aliases:
          -
          RGD1308084
          Chromosome Location:
          Chr.11: 57485628 - 57500476 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 60053698 - 60068142 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01763558_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103693563
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008768673.1 XP_008766895.1 3 - 4 543 123
          XM_008768674.1 XP_008766896.1 3 - 4 1150 123
          XM_008768675.1 XP_008766897.1 3 - 4 579 123
          Gene Symbol: Abhd10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001123352.1 NP_001116824.1 3 - 4 413 123
          XM_008768732.1 XP_008766954.1 3 - 4 1151 123
          GenBank mRNA BC088235.1 - 3 - 4 406 123
          BC161919.1 - 3 - 4 413 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          glucuronoside catabolic process
          hydrolase activity, hydrolyzing O-glycosyl compounds

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751044 - -
          Affymetrix Rat Gene 1.0 ST Array 10751045 - -
          Affymetrix Rat Gene 1.0 ST Array 10751047 - -
          Affymetrix Rat Gene 1.0 ST Array 10751046 - -
          Affymetrix Rat Gene 1.0 ST Array 10751043 - -
          Affymetrix Rat Gene 1.1 ST Array 10751018 - -
          Rat Genome 230 2.0 Array 1377022_PM_at - -
          Rat Genome 230 2.0 Array 1377022_at - -
          Affymetrix HT Rat Focus Array 1377022_at - -
          Affymetrix HT Rat Focus Array 1377022_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100002921.rn.1 TC1100000571.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002922.rn.1 TC1100000571.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002920.rn.1 TC1100000571.rn.1 RTA TC
          Rat Expression Set 230 1377022_PM_at - -
          Rat Expression Set 230 1377022_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751018 - -
          Rat Genome U34 Set rc_AA998130_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0