Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764012_g1
          See other LOC100910410 GE Assays ›
          Gene Symbol:
          LOC100910410 Lypd4
          Gene Name:
          ly6/PLAUR domain-containing protein 4-like
          Ly6/Plaur domain containing 4
          Gene Aliases:
          -
          rCG53718
          Chromosome Location:
          Chr.9: 16823205 - 16828420 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 81728552 - 81734235 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01764012_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910410
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750649.3 XP_003750697.1 4 - 5 1049 63
          Gene Symbol: Lypd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110295.1 NP_001103765.1 4 - 5 891 63
          XM_008758982.1 XP_008757204.1 4 - 5 696 63
          XM_008758983.1 XP_008757205.1 4 - 5 841 63
          XM_008758984.1 XP_008757206.1 4 - 5 691 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P958956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89057 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P958956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P958956 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P958956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719715 - -
          Affymetrix Rat Gene 1.0 ST Array 10719716 - -
          Affymetrix Rat Gene 1.0 ST Array 10719717 - -
          Affymetrix Rat Gene 1.0 ST Array 10719718 - -
          Affymetrix Rat Gene 1.1 ST Array 10719714 - -
          Affymetrix Rat Gene 1.0 ST Array 10719714 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21503245 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0