Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764760_m1
          See other FUT9 GE Assays ›
          Gene Symbol:
          Fut9
          Gene Name:
          fucosyltransferase 9 (alpha (1,3) fucosyltransferase)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 40032794 - 40237591 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01764760_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fut9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053465.1 NP_445917.1 2 - 3 332 72
          XM_008763598.1 XP_008761820.1 1 - 2 387 72
          GenBank mRNA AB049819.1 - 2 - 3 332 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002433.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AB049819.1, NM_053465.1, XM_008763598.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fut9

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          nervous system development
          fucosylation
          fucosyltransferase activity
          alpha-(1->3)-fucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P475551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150956 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168061 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475551 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475551 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P475551 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P475551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56418 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8667 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74632 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875818 - -
          Affymetrix Rat Gene 1.0 ST Array 10875820 - -
          Affymetrix Rat Gene 1.0 ST Array 10875819 - -
          Affymetrix Rat Gene 1.0 ST Array 10875822 - -
          Affymetrix Rat Gene 1.1 ST Array 10875817 - -
          Rat Genome 230 2.0 Array 1369258_PM_at - -
          Rat Genome 230 2.0 Array 1369258_at - -
          Affymetrix HT Rat Focus Array 1369258_at - -
          Affymetrix HT Rat Focus Array 1369258_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500018171.rn.1 TC0500002433.rn.1 RTA TC
          Rat Expression Set 230 1369258_PM_at - -
          Rat Expression Set 230 1369258_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875817 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21742723 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363444 - -
          RatRef-12 Expression BeadChip ILMN_67028 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5968f498c6-wvnc9:80/100.66.128.131:80.
          git-commit: b7f02e98b2654af9c52ff750aebd516a8b607fe3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/PDP_Cmrc_Card_Notification_Changes/CFGMAGJ/CFGMAGJ-5102/CFGMAGJ-5166-COMMERCE-CARD-TRANSLATION_APAC