Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765446_g1
          See other NEWGENE_1562258 GE Assays ›
          Gene Symbol:
          NEWGENE_1562258 LOC103692173 Wbp1
          Gene Name:
          INO80 complex subunit B
          WW domain-binding protein 1
          WW domain binding protein 1
          Gene Aliases:
          Ino80b
          -
          -
          Chromosome Location:
          Chr.4: 114817603 - 114823889 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 114817601 - 114819959 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 113951873 - 113954272 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01765446_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEWGENE_1562258
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763087.1 XP_008761309.1 7 - 8 2729 86
          XM_008763089.1 XP_008761311.1 7 - 8 2967 86
          Gene Symbol: LOC103692173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763090.1 XP_008761312.1 4 - 5 543 86
          XM_008763091.1 XP_008761313.1 4 - 5 471 86
          XM_008763092.1 XP_008761314.1 3 - 4 462 86
          XM_008763093.1 XP_008761315.1 3 - 4 364 86
          Gene Symbol: Wbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106600.1 NP_001100070.1 503 86
          XM_006236712.2 XP_006236774.1 562 86
          XM_006236713.2 XP_006236775.1 471 86
          XM_008763021.1 XP_008761243.1 364 86
          GenBank mRNA BC079453.1 - 1060 86
          BC098709.1 - 3 - 4 1057 86
          BC158831.1 - 460 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004158.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P474158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533960 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P474158 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P474158 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P474158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863494 - -
          Affymetrix Rat Gene 1.0 ST Array 10863492 - -
          Affymetrix Rat Gene 1.0 ST Array 10863493 - -
          Affymetrix Rat Gene 1.0 ST Array 10863491 - -
          Affymetrix Rat Gene 1.0 ST Array 10863490 - -
          Affymetrix Rat Gene 1.1 ST Array 10863489 - -
          Rat Genome 230 2.0 Array 1380108_PM_at - -
          Rat Genome 230 2.0 Array 1372232_PM_at - -
          Rat Genome 230 2.0 Array 1372232_at - -
          Rat Genome 230 2.0 Array 1374450_PM_at - -
          Rat Genome 230 2.0 Array 1380108_at - -
          Rat Genome 230 2.0 Array 1374450_at - -
          Affymetrix HT Rat Focus Array 1372232_PM_at - -
          Affymetrix HT Rat Focus Array 1380108_at - -
          Affymetrix HT Rat Focus Array 1372232_at - -
          Affymetrix HT Rat Focus Array 1380108_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400029817.rn.1 TC0400004158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029818.rn.1 TC0400004158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400029819.rn.1 TC0400004158.rn.1 RTA TC
          Rat Expression Set 230 1372232_PM_at - -
          Rat Expression Set 230 1372232_at - -
          Rat Expression Set 230 1380108_at - -
          Rat Expression Set 230 1380108_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863489 - -
          Rat Genome U34 Set rc_AI170808_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21117403 - -
          ABI Rat Genome Survey Microarray 21998548 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373448 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0