Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766343_g1
          See other LOC102555476 GE Assays ›
          Gene Symbol:
          LOC102555476 Cc2d2b
          Gene Name:
          uncharacterized LOC102555476
          coiled-coil and C2 domain containing 2B
          Gene Aliases:
          -
          RGD1310408
          Chromosome Location:
          Chr.1: 259828574 - 259876410 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 259998716 - 260046366 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01766343_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102555476
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760504.1 XP_008758726.1 7 - 8 638 64
          Gene Symbol: Cc2d2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760505.1 XP_008758727.1 638 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144373 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144373 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144373 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P316410 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715286 - -
          Affymetrix Rat Gene 1.0 ST Array 10715299 - -
          Affymetrix Rat Gene 1.0 ST Array 10715280 - -
          Affymetrix Rat Gene 1.0 ST Array 10715297 - -
          Affymetrix Rat Gene 1.0 ST Array 10715273 - -
          Affymetrix Rat Gene 1.0 ST Array 10715294 - -
          Affymetrix Rat Gene 1.0 ST Array 10715278 - -
          Affymetrix Rat Gene 1.0 ST Array 10715283 - -
          Affymetrix Rat Gene 1.0 ST Array 10715291 - -
          Affymetrix Rat Gene 1.0 ST Array 10715287 - -
          Affymetrix Rat Gene 1.0 ST Array 10715274 - -
          Affymetrix Rat Gene 1.0 ST Array 10715295 - -
          Affymetrix Rat Gene 1.0 ST Array 10715288 - -
          Affymetrix Rat Gene 1.0 ST Array 10715300 - -
          Affymetrix Rat Gene 1.0 ST Array 10715275 - -
          Affymetrix Rat Gene 1.0 ST Array 10715289 - -
          Affymetrix Rat Gene 1.0 ST Array 10715293 - -
          Affymetrix Rat Gene 1.0 ST Array 10715285 - -
          Affymetrix Rat Gene 1.0 ST Array 10715276 - -
          Affymetrix Rat Gene 1.0 ST Array 10715281 - -
          Affymetrix Rat Gene 1.0 ST Array 10715298 - -
          Affymetrix Rat Gene 1.0 ST Array 10715269 - -
          Affymetrix Rat Gene 1.0 ST Array 10715296 - -
          Affymetrix Rat Gene 1.0 ST Array 10715271 - -
          Affymetrix Rat Gene 1.0 ST Array 10715290 - -
          Affymetrix Rat Gene 1.0 ST Array 10715292 - -
          Affymetrix Rat Gene 1.0 ST Array 10715282 - -
          Affymetrix Rat Gene 1.0 ST Array 10715279 - -
          Affymetrix Rat Gene 1.0 ST Array 10715270 - -
          Affymetrix Rat Gene 1.0 ST Array 10715268 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362807 - -
          RatRef-12 Expression BeadChip ILMN_55828 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0