Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766377_m1
          See other ZFP518A GE Assays ›
          Gene Symbol:
          Zfp518a
          Gene Name:
          zinc finger protein 518A
          Gene Aliases:
          RGD1305314, Znf518, Znf518a
          Chromosome Location:
          Chr.1: 260152893 - 260178360 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766377_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp518a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030038.1 NP_001025209.1 2 - 3 132 68
          XM_006231397.2 XP_006231459.1 2 - 3 309 68
          XM_006231398.2 XP_006231460.1 2 - 3 349 68
          XM_008760440.1 XP_008758662.1 2 - 3 445 68
          GenBank mRNA BC099801.1 - 2 - 3 132 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003609.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC099801.1, NM_001030038.1, XM_006231397.2, XM_006231398.2, XM_008760440.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp518a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99144 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143675 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715315 - -
          Affymetrix Rat Gene 1.0 ST Array 10715311 - -
          Affymetrix Rat Gene 1.0 ST Array 10715312 - -
          Affymetrix Rat Gene 1.0 ST Array 10715313 - -
          Affymetrix Rat Gene 1.0 ST Array 10715314 - -
          Affymetrix Rat Gene 1.1 ST Array 10715310 - -
          Rat Genome 230 2.0 Array 1384940_at - -
          Rat Genome 230 2.0 Array 1384940_PM_at - -
          Affymetrix HT Rat Focus Array 1384940_at - -
          Affymetrix HT Rat Focus Array 1384940_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100029399.rn.1 TC0100003609.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029398.rn.1 TC0100003609.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029397.rn.1 TC0100003609.rn.1 RTA TC
          Rat Expression Set 230 1384940_at - -
          Rat Expression Set 230 1384940_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715310 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21974961 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375232 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0