Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767467_m1
          See other ING2 GE Assays ›
          Gene Symbol:
          Ing2
          Gene Name:
          inhibitor of growth family, member 2
          Gene Aliases:
          Ing1l
          Chromosome Location:
          Chr.16: 47714999 - 47725836 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767467_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ing2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106083.1 NP_001099553.1 1 - 2 607 139
          GenBank mRNA BC168652.1 - 587 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000546.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          regulation of transcription, DNA-templated
          male meiosis I
          spermatogenesis
          spermatid development
          biological_process
          negative regulation of cell population proliferation
          flagellated sperm motility
          positive regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of apoptotic process
          positive regulation of transcription, DNA-templated
          male germ-line stem cell asymmetric division
          seminiferous tubule development
          negative regulation of apoptotic signaling pathway
          molecular_function
          DNA binding
          chromatin binding
          zinc ion binding
          methylated histone binding
          phosphatidylinositol binding
          protein-containing complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P293008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P293008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P293008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P293008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P293008 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788136 - -
          Affymetrix Rat Gene 1.0 ST Array 10788135 - -
          Affymetrix Rat Gene 1.1 ST Array 10788134 - -
          Rat Genome 230 2.0 Array 1392165_PM_at - -
          Rat Genome 230 2.0 Array 1397243_at - -
          Rat Genome 230 2.0 Array 1386129_PM_at - -
          Rat Genome 230 2.0 Array 1392165_at - -
          Rat Genome 230 2.0 Array 1386129_at - -
          Rat Genome 230 2.0 Array 1397243_PM_at - -
          Affymetrix HT Rat Focus Array 1386129_PM_at - -
          Affymetrix HT Rat Focus Array 1392165_at - -
          Affymetrix HT Rat Focus Array 1386129_at - -
          Affymetrix HT Rat Focus Array 1392165_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600004446.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004450.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004441.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004440.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004447.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004449.rn.1 TC1600000546.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004448.rn.1 TC1600000546.rn.1 RTA TC
          Rat Expression Set 230 1386129_at - -
          Rat Expression Set 230 1397243_PM_at - -
          Rat Expression Set 230 1392165_at - -
          Rat Expression Set 230 1392165_PM_at - -
          Rat Expression Set 230 1386129_PM_at - -
          Rat Expression Set 230 1397243_at - -
          Affymetrix Rat Gene 1.0 ST Array 10788134 - -
          Rat Genome U34 Set rc_AI071247_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21320661 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365602 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0