Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771058_m1
          See other GPR143 GE Assays ›
          Gene Symbol:
          Gpr143
          Gene Name:
          G protein-coupled receptor 143
          Gene Aliases:
          RGD1565799
          Chromosome Location:
          Chr.X: 23668363 - 23693162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771058_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr143
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106958.1 NP_001100428.1 8 - 9 1191 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000251.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          neuropeptide signaling pathway
          melanosome localization
          melanosome transport
          melanosome organization
          calcium-mediated signaling using intracellular calcium source
          phosphatidylinositol-mediated signaling
          regulation of calcium-mediated signaling
          G protein-coupled receptor activity
          dopamine binding
          L-DOPA receptor activity
          L-DOPA binding
          tyrosine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146076 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071810 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P534536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933222 - -
          Affymetrix Rat Gene 1.0 ST Array 10933217 - -
          Affymetrix Rat Gene 1.0 ST Array 10933221 - -
          Affymetrix Rat Gene 1.0 ST Array 10933223 - -
          Affymetrix Rat Gene 1.0 ST Array 10933219 - -
          Affymetrix Rat Gene 1.0 ST Array 10933220 - -
          Affymetrix Rat Gene 1.0 ST Array 10933226 - -
          Affymetrix Rat Gene 1.0 ST Array 10933218 - -
          Affymetrix Rat Gene 1.0 ST Array 10933224 - -
          Affymetrix Rat Gene 1.0 ST Array 10933225 - -
          Affymetrix Rat Gene 1.1 ST Array 10933216 - -
          Rat Genome 230 2.0 Array 1385419_PM_at - -
          Rat Genome 230 2.0 Array 1385419_at - -
          Affymetrix HT Rat Focus Array 1385419_PM_at - -
          Affymetrix HT Rat Focus Array 1385419_at - -
          Affymetrix Rat Clariom S Assay PSR0X00001902.rn.1 TC0X00000251.rn.1 RTA TC
          Rat Expression Set 230 1385419_at - -
          Rat Expression Set 230 1385419_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933216 - -
          Rat Genome U34 Set rc_AI073065_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21608809 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0