Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01772640_m1
          See other LOC691813 GE Assays ›
          Gene Symbol:
          LOC691813 Pla2g4c
          Gene Name:
          similar to phospholipase A2, group IVC (cytosolic, calcium-independent)
          phospholipase A2, group IVC (cytosolic, calcium-independent)
          Gene Aliases:
          -
          Pla2g4cl1
          Chromosome Location:
          Chr.1: 75495214 - 75580892 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 75440035 - 75481995 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01772640_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC691813
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006228335.2 XP_006228397.1 725 89
          XM_008759023.1 XP_008757245.1 725 89
          XM_008759024.1 XP_008757246.1 721 89
          Gene Symbol: Pla2g4c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145982.1 NP_001139454.1 3 - 4 443 89
          GenBank mRNA AB113213.1 - 3 - 4 428 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000899.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid catabolic process
          glycerophospholipid catabolic process
          phospholipase activity
          calcium-independent phospholipase A2 activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P070313 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44698 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P070338 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084723 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P070338 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084723 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440395 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704347 - -
          Affymetrix Rat Gene 1.0 ST Array 10704344 - -
          Affymetrix Rat Gene 1.0 ST Array 10704345 - -
          Affymetrix Rat Gene 1.0 ST Array 10704348 - -
          Affymetrix Rat Gene 1.1 ST Array 10704341 - -
          Affymetrix Rat Clariom S Assay PSR0100006109.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006114.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006108.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006113.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006110.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006111.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006115.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006112.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006116.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006107.rn.1 TC0100000899.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006117.rn.1 TC0100000899.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22129768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0