Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01774859_g1
          See other LOC103689933 GE Assays ›
          Gene Symbol:
          LOC103689933 Klk1c3
          Gene Name:
          glandular kallikrein-3, submandibular
          kallikrein 1-related peptidase C3
          Gene Aliases:
          -
          KLK3, Klk1c10l2, RSGK-50, RSKG-50, rGK-3
          Chromosome Location:
          Chr.1: 100160127 - 100163941 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 100164399 - 100165474 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01774859_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103689933
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008759445.1 XP_008757667.1 4 - 5 645 165
          Gene Symbol: Klk1c3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271315.1 NP_001258244.1 606 165
          GenBank mRNA L33840.1 - 558 165
          M11564.1 - 165

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001437.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P374824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P374824 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P374824 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374823 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374824 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706497 - -
          Affymetrix Rat Gene 1.0 ST Array 10706496 - -
          Affymetrix Rat Gene 1.1 ST Array 10706492 - -
          Rat Genome 230 2.0 Array 1371084_PM_at - -
          Rat Genome 230 2.0 Array 1371084_at - -
          Affymetrix HT Rat Focus Array 1371084_PM_at - -
          Affymetrix HT Rat Focus Array 1371084_at - -
          Affymetrix Rat Clariom S Assay PSR0100011054.rn.1 TC0100001437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100011053.rn.1 TC0100001437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100011052.rn.1 TC0100001437.rn.1 RTA TC
          Rat Expression Set 230 1371084_at - -
          Rat Expression Set 230 1371084_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706492 - -
          Rat Genome U34 Set M26534_f_at - -
          Rat Genome U34 Set M26534_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22376816 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0