Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01786196_g1
          See other RT1-DMA GE Assays ›
          Gene Symbol:
          RT1-DMa LOC100909593
          Gene Name:
          RT1 class II, locus DMa
          class II histocompatibility antigen, M alpha chain-like
          Gene Aliases:
          DMA1, RT1.DMa, RT1.Ma
          -
          Chromosome Location:
          Chr.20: 5240975 - 5244378 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 3935501 - 3938915 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01786196_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RT1-DMa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198741.2 NP_942036.1 4 - 5 835 93
          GenBank mRNA BC091107.1 - 4 - 5 818 93
          Z49761.1 - 4 - 5 803 93
          Gene Symbol: LOC100909593
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006256035.2 XP_006256097.1 4 - 5 846 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RT1-DMa
          Click here to find siRNAs for Gene LOC100909593

          Gene Ontology Categories:

          Function(s) Process(es)

          peptide antigen assembly with MHC class II protein complex
          antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          positive regulation of germinal center formation
          positive regulation of humoral immune response
          immune response
          protein transport
          immunoglobulin mediated immune response
          antigen processing and presentation
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          positive thymic T cell selection
          positive regulation of T cell differentiation
          inner ear development
          positive regulation of immune response
          chaperone cofactor-dependent protein refolding
          protein-containing complex assembly
          MHC class II protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154250 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128478 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128478 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P473314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831630 - -
          Affymetrix Rat Gene 1.0 ST Array 10831632 - -
          Affymetrix Rat Gene 1.0 ST Array 10831629 - -
          Affymetrix Rat Gene 1.0 ST Array 10831631 - -
          Affymetrix Rat Gene 1.0 ST Array 10831633 - -
          Affymetrix Rat Gene 1.1 ST Array 10831628 - -
          Rat Genome 230 2.0 Array 1370904_PM_at - -
          Rat Genome 230 2.0 Array 1370904_at - -
          Affymetrix HT Rat Focus Array 1370904_at - -
          Affymetrix HT Rat Focus Array 1370904_PM_at - -
          Rat Expression Set 230 1370904_at - -
          Rat Expression Set 230 1370904_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831628 - -
          Rat Genome U34 Set U31598_s_at - -
          Rat Genome U34 Set Z49761_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21123962 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039346 - -
          RatRef-12 Expression BeadChip ILMN_50678 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0