Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01786262_m1
          See other NAPEPLD GE Assays ›
          Gene Symbol:
          Napepld
          Gene Name:
          N-acyl phosphatidylethanolamine phospholipase D
          Gene Aliases:
          NAPE-PLD
          Chromosome Location:
          Chr.4: 9965324 - 10004650 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01786262_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Napepld
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199381.1 NP_955413.1 3 - 4 1054 71
          XM_006235908.2 XP_006235970.1 4 - 5 2476 71
          XM_006235910.2 XP_006235972.1 4 - 5 1323 71
          XM_006235911.2 XP_006235973.1 4 - 5 1277 71
          XM_008762638.1 XP_008760860.1 4 - 5 4725 71
          XM_008762639.1 XP_008760861.1 4 - 5 1986 71
          GenBank mRNA AB112351.1 - 3 - 4 1054 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000158.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Napepld

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid metabolic process
          phospholipid catabolic process
          phospholipase activity
          zinc ion binding
          N-acylphosphatidylethanolamine-specific phospholipase D activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148535 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P166489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2939 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148535 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P166489 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853133 - -
          Affymetrix Rat Gene 1.0 ST Array 10853135 - -
          Affymetrix Rat Gene 1.0 ST Array 10853136 - -
          Affymetrix Rat Gene 1.0 ST Array 10853134 - -
          Affymetrix Rat Gene 1.1 ST Array 10853132 - -
          Affymetrix Rat Clariom S Assay PSR0400001224.rn.1 TC0400000158.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10853132 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21402931 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373161 - -
          RatRef-12 Expression BeadChip ILMN_50245 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0