Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02106015_u1
          See other ITGB8 GE Assays ›
          Gene Symbol:
          Itgb8
          Gene Name:
          integrin, beta 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 147089280 - 147172845 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02106015_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108726.1 NP_001102196.1 11 2523 141
          XM_006240697.2 XP_006240759.1 13 3057 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003507.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside metabolic process
          cell-matrix adhesion
          integrin-mediated signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          cartilage development
          signaling receptor binding
          signaling receptor activity
          extracellular matrix protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P055684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055684 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P462029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75912 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892845 - -
          Affymetrix Rat Gene 1.0 ST Array 10892843 - -
          Affymetrix Rat Gene 1.0 ST Array 10892847 - -
          Affymetrix Rat Gene 1.0 ST Array 10892846 - -
          Affymetrix Rat Gene 1.0 ST Array 10892848 - -
          Affymetrix Rat Gene 1.0 ST Array 10892837 - -
          Affymetrix Rat Gene 1.0 ST Array 10892838 - -
          Affymetrix Rat Gene 1.0 ST Array 10892840 - -
          Affymetrix Rat Gene 1.0 ST Array 10892839 - -
          Affymetrix Rat Gene 1.0 ST Array 10892836 - -
          Affymetrix Rat Gene 1.0 ST Array 10892844 - -
          Affymetrix Rat Gene 1.1 ST Array 10892835 - -
          Rat Genome 230 2.0 Array 1377974_PM_at - -
          Rat Genome 230 2.0 Array 1377974_at - -
          Rat Genome 230 2.0 Array 1395372_at - -
          Rat Genome 230 2.0 Array 1395372_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600024856.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024857.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024859.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024862.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024865.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024867.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024868.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024874.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024863.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024872.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024855.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024864.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024860.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024866.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600024873.rn.1 TC0600003507.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10892835 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21652028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353484 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0