Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126273.1 | NP_001119745.1 | 3 | 1130 | 105 | |
XM_006249498.2 | XP_006249560.1 | 4 | 869 | 105 | ||
GenBank mRNA | BC166547.1 | - | 3 | 1130 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |