Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02106611_u1
          See other TSC22D4 GE Assays ›
          Gene Symbol:
          Tsc22d4 LOC102550456
          Gene Name:
          TSC22 domain family, member 4
          TSC22 domain family protein 4-like
          Gene Aliases:
          Tilz2, thg-1pit
          -
          Chromosome Location:
          Chr.12: 22000566 - 22021860 on Build Rnor_6.0
          Chromosome Location:
          Chr.12: 21748608 - 21761501 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02106611_u1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsc22d4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006249186.2 XP_006249248.1 3 1651 144
          GenBank mRNA BC105911.1 - 2 872 144
          FM049873.1 - 1 102 144
          Gene Symbol: LOC102550456
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006249077.2 XP_006249139.1 1646 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001892.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          response to osmotic stress
          DNA-binding transcription factor activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P777424 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153239 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140081 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140081 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140081 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P777424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757413 - -
          Affymetrix Rat Gene 1.0 ST Array 10757418 - -
          Affymetrix Rat Gene 1.0 ST Array 10757411 - -
          Affymetrix Rat Gene 1.0 ST Array 10757410 - -
          Affymetrix Rat Gene 1.0 ST Array 10757416 - -
          Affymetrix Rat Gene 1.0 ST Array 10757417 - -
          Affymetrix Rat Gene 1.0 ST Array 10757412 - -
          Affymetrix Rat Gene 1.0 ST Array 10757419 - -
          Affymetrix Rat Gene 1.0 ST Array 10757414 - -
          Affymetrix Rat Gene 1.1 ST Array 10757409 - -
          Affymetrix Rat Gene 1.1 ST Array 10757415 - -
          Rat Genome 230 2.0 Array 1371550_PM_at - -
          Rat Genome 230 2.0 Array 1375958_at - -
          Rat Genome 230 2.0 Array 1377159_at - -
          Rat Genome 230 2.0 Array 1375958_PM_at - -
          Rat Genome 230 2.0 Array 1377159_PM_at - -
          Rat Genome 230 2.0 Array 1371550_at - -
          Rat Genome 230 2.0 Array 1382315_PM_at - -
          Rat Genome 230 2.0 Array 1382315_at - -
          Affymetrix HT Rat Focus Array 1371550_PM_at - -
          Affymetrix HT Rat Focus Array 1382315_PM_at - -
          Affymetrix HT Rat Focus Array 1382315_at - -
          Affymetrix HT Rat Focus Array 1371550_at - -
          Affymetrix Rat Clariom S Assay PSR1200016422.rn.1 TC1200001892.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200016431.rn.1 TC1200001892.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200016433.rn.1 TC1200001892.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200016425.rn.1 TC1200001892.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200016423.rn.1 TC1200001892.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200016421.rn.1 TC1200001892.rn.1 RTA TC
          Rat Expression Set 230 1377159_at - -
          Rat Expression Set 230 1377159_PM_at - -
          Rat Expression Set 230 1371550_PM_at - -
          Rat Expression Set 230 1375958_PM_at - -
          Rat Expression Set 230 1371550_at - -
          Rat Expression Set 230 1375958_at - -
          Rat Expression Set 230 1382315_at - -
          Rat Expression Set 230 1382315_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757415 - -
          Affymetrix Rat Gene 1.0 ST Array 10757409 - -
          Rat Genome U34 Set rc_AA944353_at - -
          Rat Genome U34 Set rc_AI044932_at - -
          Rat Genome U34 Set rc_AI229113_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix