Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256487.2 | XP_006256549.1 | 11 | 1703 | 124 | |
XM_008772957.1 | XP_008771179.1 | 13 | 3126 | 124 | ||
XM_008772958.1 | XP_008771180.1 | 13 | 3126 | 124 | ||
XM_008772959.1 | XP_008771181.1 | 12 | 2935 | 124 | ||
XM_008772961.1 | XP_008771183.1 | 9 | 1329 | 124 | ||
XM_574753.6 | XP_574753.4 | 10 | 1507 | 124 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |