Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02112097_g1
          See other RPS6 GE Assays ›
          Gene Symbol:
          Rps6 LOC100911372
          Gene Name:
          ribosomal protein S6
          40S ribosomal protein S6-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.5: 105197821 - 105200681 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 105209844 - 105212791 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02112097_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017160.1 NP_058856.1 682 163
          GenBank mRNA BC058149.1 - 712 163
          FQ211324.1 - 314 163
          FQ217720.1 - 314 163
          FQ218111.1 - 314 163
          FQ229168.1 - 314 163
          FQ230362.1 - 314 163
          FQ231606.1 - 314 163
          M29358.1 - 682 163
          Gene Symbol: LOC100911372
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749971.3 XP_003750019.1 5 767 163

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps6
          Click here to find siRNAs for Gene LOC100911372

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          G1/S transition of mitotic cell cycle
          mitotic cell cycle
          placenta development
          T cell proliferation involved in immune response
          rRNA processing
          translation
          activation-induced cell death of T cells
          mitotic cell cycle checkpoint signaling
          gastrulation
          mammalian oogenesis stage
          TOR signaling
          T cell differentiation in thymus
          ribosomal small subunit biogenesis
          glucose homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          erythrocyte development
          RNA binding
          structural constituent of ribosome
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112093 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85044 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045330 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045330 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045330 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112093 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877920 - -
          Affymetrix Rat Gene 1.0 ST Array 10877917 - -
          Affymetrix Rat Gene 1.0 ST Array 10877919 - -
          Affymetrix Rat Gene 1.0 ST Array 10877918 - -
          Affymetrix Rat Gene 1.1 ST Array 10877916 - -
          Rat Genome 230 2.0 Array 1367573_at - -
          Rat Genome 230 2.0 Array 1367573_PM_at - -
          Affymetrix HT Rat Focus Array 1367573_PM_at - -
          Affymetrix HT Rat Focus Array 1367573_at - -
          Rat Expression Set 230 1367573_PM_at - -
          Rat Expression Set 230 1367573_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877916 - -
          Rat Genome U34 Set M29358_g_at - -
          Rat Genome U34 Set M29358_at - -
          Rat Genome U34 Set rc_AI178582_s_at - -
          Rat Genome U34 Set rc_AI136296_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22174155 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364372 - -
          RatRef-12 Expression BeadChip ILMN_68946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0