Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02112959_s1
          See other C1D GE Assays ›
          Gene Symbol:
          C1d
          Gene Name:
          C1D nuclear receptor co-repressor
          Gene Aliases:
          RGD1560600
          Chromosome Location:
          Chr.14: 100415645 - 100427878 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02112959_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C1d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106021.1 NP_001099491.1 5 3068 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001055.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          maturation of 5.8S rRNA
          negative regulation of transcription, DNA-templated
          DNA binding
          transcription corepressor activity
          RNA binding
          nuclear receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P077085 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P716856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P716856 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093382 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093382 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P716856 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P457008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14774 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774335 - -
          Affymetrix Rat Gene 1.0 ST Array 10774332 - -
          Affymetrix Rat Gene 1.0 ST Array 10774333 - -
          Affymetrix Rat Gene 1.0 ST Array 10774334 - -
          Affymetrix Rat Gene 1.0 ST Array 10774336 - -
          Affymetrix Rat Gene 1.1 ST Array 10774331 - -
          Rat Genome 230 2.0 Array 1377731_at - -
          Rat Genome 230 2.0 Array 1397387_PM_at - -
          Rat Genome 230 2.0 Array 1397387_at - -
          Rat Genome 230 2.0 Array 1377731_PM_at - -
          Affymetrix HT Rat Focus Array 1377731_at - -
          Affymetrix HT Rat Focus Array 1377731_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400007699.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007707.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007706.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007705.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007711.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007709.rn.1 TC1400001055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400007704.rn.1 TC1400001055.rn.1 RTA TC
          Rat Expression Set 230 1377731_PM_at - -
          Rat Expression Set 230 1377731_at - -
          Rat Expression Set 230 1397387_PM_at - -
          Rat Expression Set 230 1397387_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774331 - -
          Rat Genome U34 Set rc_AA891969_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21182395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0