Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347093_m1
          See other APOH GE Assays ›
          Gene Symbol:
          Apoh
          Gene Name:
          apolipoprotein H (beta-2-glycoprotein I)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 96640013 - 96653939 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347093_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009626.1 NP_001009626.1 3 - 4 350 132
          GenBank mRNA AM071386.1 - 3 - 4 309 132
          BC088180.1 - 3 - 4 350 132
          FQ209412.1 - 3 - 4 389 132
          FQ209491.1 - 3 - 4 376 132
          FQ209740.1 - 3 - 4 385 132
          FQ210182.1 - 3 - 4 376 132
          FQ210311.1 - 3 - 4 376 132
          FQ210756.1 - 3 - 4 392 132
          FQ210813.1 - 3 - 4 376 132
          FQ211328.1 - 3 - 4 349 132
          FQ218445.1 - 3 - 4 390 132
          FQ218454.1 - 3 - 4 376 133
          FQ218597.1 - 3 - 4 372 132
          FQ218599.1 - 3 - 4 376 132
          FQ218637.1 - 3 - 4 376 132
          FQ218859.1 - 3 - 4 349 132
          FQ218865.1 - 3 - 4 376 132
          FQ219138.1 - 3 - 4 441 132
          FQ219328.1 - 3 - 4 376 132
          FQ219380.1 - 3 - 4 377 132
          FQ219438.1 - 3 - 4 375 132
          FQ219481.1 - 3 - 4 389 132
          FQ219537.1 - 3 - 4 373 132
          FQ219580.1 - 3 - 4 390 132
          FQ219601.1 - 3 - 4 389 132
          FQ219674.1 - 3 - 4 376 132
          X15551.1 - 3 - 4 205 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001835.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoh

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          triglyceride metabolic process
          blood coagulation
          blood coagulation, intrinsic pathway
          negative regulation of endothelial cell migration
          negative regulation of angiogenesis
          regulation of blood coagulation
          negative regulation of blood coagulation
          animal organ regeneration
          plasminogen activation
          negative regulation of myeloid cell apoptotic process
          triglyceride transport
          negative regulation of smooth muscle cell apoptotic process
          positive regulation of lipoprotein lipase activity
          regulation of fibrinolysis
          negative regulation of fibrinolysis
          protein binding
          phospholipid binding
          heparin binding
          lipid binding
          identical protein binding
          lipoprotein lipase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1000016631.rn.1 TC1000001835.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0