Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347357_m1
          See other WBP11L1 GE Assays ›
          Gene Symbol:
          Wbp11l1 Wbp11
          Gene Name:
          WW domain-binding protein 11-like 1
          WW domain binding protein 11
          Gene Aliases:
          -
          wbp-11
          Chromosome Location:
          Chr.4: 170154259 - 170167722 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 170772164 - 170785627 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02347357_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wbp11l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763449.1 XP_008761671.1 1096 133
          XM_008763450.1 XP_008761672.1 1098 133
          Gene Symbol: Wbp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008763375.1 8 - 9 1096 133
          GenBank mRNA BC087082.1 - 8 - 9 1009 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003956.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          mRNA processing
          RNA splicing
          regulation of catalytic activity
          RNA binding
          protein phosphatase regulator activity
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P666259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1040 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P666259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P666259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P666259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P544297 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P666259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101874 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866494 - -
          Affymetrix Rat Gene 1.0 ST Array 10866491 - -
          Affymetrix Rat Gene 1.0 ST Array 10866496 - -
          Affymetrix Rat Gene 1.0 ST Array 10866498 - -
          Affymetrix Rat Gene 1.0 ST Array 10866490 - -
          Affymetrix Rat Gene 1.0 ST Array 10866497 - -
          Affymetrix Rat Gene 1.0 ST Array 10866495 - -
          Affymetrix Rat Gene 1.0 ST Array 10891391 - -
          Affymetrix Rat Gene 1.0 ST Array 10866499 - -
          Affymetrix Rat Gene 1.0 ST Array 10866493 - -
          Affymetrix Rat Gene 1.0 ST Array 10866489 - -
          Affymetrix Rat Gene 1.0 ST Array 10866492 - -
          Affymetrix Rat Gene 1.1 ST Array 10782647 - -
          Affymetrix Rat Gene 1.1 ST Array 10866488 - -
          Affymetrix Rat Gene 1.1 ST Array 10891390 - -
          Rat Genome 230 2.0 Array 1372575_at - -
          Rat Genome 230 2.0 Array 1372575_PM_at - -
          Affymetrix HT Rat Focus Array 1372575_at - -
          Affymetrix HT Rat Focus Array 1372575_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400028066.rn.1 TC0400003956.rn.1 RTA TC
          Rat Expression Set 230 1372575_PM_at - -
          Rat Expression Set 230 1372575_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866488 - -
          Rat Genome U34 Set rc_AA892364_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371715 - -
          RatRef-12 Expression BeadChip ILMN_52064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0