Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347924_m1
          See other KCNC3 GE Assays ›
          Gene Symbol:
          Kcnc3
          Gene Name:
          potassium channel, voltage gated Shaw-related subfamily C, member 3
          Gene Aliases:
          KShIIID, Kv3.3
          Chromosome Location:
          Chr.1: 100593476 - 100607874 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347924_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053997.4 NP_446449.2 3 - 4 2393 88
          XM_006228965.2 XP_006229027.1 4 - 5 2620 88
          XM_006228967.2 XP_006229029.1 3 - 4 2590 88
          GenBank mRNA AY179603.1 - 3 - 4 2393 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001459.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnc3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          regulation of neurotransmitter secretion
          protein homooligomerization
          potassium ion transmembrane transport
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P220139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21538 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081352 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081357 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081352 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081357 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081352 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081357 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007439 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384086 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P640175 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P104687 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702138249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26012 - -
          Agilent Rat cDNA Microarray (G4105A) 702138249 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706611 - -
          Affymetrix Rat Gene 1.0 ST Array 10706602 - -
          Affymetrix Rat Gene 1.0 ST Array 10706607 - -
          Affymetrix Rat Gene 1.0 ST Array 10706605 - -
          Affymetrix Rat Gene 1.0 ST Array 10706608 - -
          Affymetrix Rat Gene 1.0 ST Array 10706604 - -
          Affymetrix Rat Gene 1.0 ST Array 10706603 - -
          Affymetrix Rat Gene 1.0 ST Array 10706609 - -
          Affymetrix Rat Gene 1.0 ST Array 10706610 - -
          Affymetrix Rat Gene 1.0 ST Array 10706606 - -
          Affymetrix Rat Gene 1.0 ST Array 10706601 - -
          Affymetrix Rat Gene 1.0 ST Array 10706599 - -
          Affymetrix Rat Gene 1.0 ST Array 10706598 - -
          Affymetrix Rat Gene 1.1 ST Array 10706597 - -
          Rat Genome 230 2.0 Array 1388162_PM_a_at - -
          Rat Genome 230 2.0 Array 1388162_a_at - -
          Rat Genome 230 2.0 Array 1369133_a_at - -
          Rat Genome 230 2.0 Array 1369134_PM_x_at - -
          Rat Genome 230 2.0 Array 1369134_x_at - -
          Rat Genome 230 2.0 Array 1370958_at - -
          Rat Genome 230 2.0 Array 1369133_PM_a_at - -
          Rat Genome 230 2.0 Array 1389120_PM_at - -
          Rat Genome 230 2.0 Array 1370958_PM_at - -
          Rat Genome 230 2.0 Array 1389120_at - -
          Affymetrix HT Rat Focus Array 1370958_PM_at - -
          Affymetrix HT Rat Focus Array 1369133_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369134_PM_x_at - -
          Affymetrix HT Rat Focus Array 1388162_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369134_x_at - -
          Affymetrix HT Rat Focus Array 1388162_a_at - -
          Affymetrix HT Rat Focus Array 1370958_at - -
          Affymetrix HT Rat Focus Array 1369133_a_at - -
          Affymetrix HT Rat Focus Array 1389120_at - -
          Affymetrix HT Rat Focus Array 1389120_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100011308.rn.1 TC0100001459.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100011309.rn.1 TC0100001459.rn.1 RTA TC
          Rat Expression Set 230 1369133_PM_a_at - -
          Rat Expression Set 230 1369134_PM_x_at - -
          Rat Expression Set 230 1388162_PM_a_at - -
          Rat Expression Set 230 1369133_a_at - -
          Rat Expression Set 230 1388162_a_at - -
          Rat Expression Set 230 1389120_at - -
          Rat Expression Set 230 1389120_PM_at - -
          Rat Expression Set 230 1370958_at - -
          Rat Expression Set 230 1370958_PM_at - -
          Rat Expression Set 230 1369134_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706597 - -
          Rat Genome U34 Set rc_AI045315_f_at - -
          Rat Genome U34 Set rc_AI639023_at - -
          Rat Genome U34 Set M84210_at - -
          Rat Genome U34 Set rc_AI007939_f_at - -
          Rat Genome U34 Set M84210_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368808 - -
          RatRef-12 Expression BeadChip ILMN_61403 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0