Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02347948_m1
          See other ARF3 GE Assays ›
          Gene Symbol:
          Arf3
          Gene Name:
          ADP-ribosylation factor 3
          Gene Aliases:
          Ac1-253
          Chromosome Location:
          Chr.7: 140412468 - 140438403 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02347948_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080904.2 NP_543180.1 1 - 2 225 128
          XM_006257320.2 XP_006257382.1 2 - 3 434 128
          GenBank mRNA AY325223.1 - 2 - 3 192 128
          BC088865.1 - 1 - 2 225 128
          L12382.1 - 2 - 3 338 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arf3

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090131 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P683655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P683655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P683655 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P683655 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P090131 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P683655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17815 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906945 - -
          Affymetrix Rat Gene 1.0 ST Array 10906948 - -
          Affymetrix Rat Gene 1.0 ST Array 10906946 - -
          Affymetrix Rat Gene 1.0 ST Array 10906947 - -
          Affymetrix Rat Gene 1.0 ST Array 10906949 - -
          Affymetrix Rat Gene 1.1 ST Array 10906944 - -
          Rat Genome 230 2.0 Array 1388997_PM_at - -
          Rat Genome 230 2.0 Array 1388997_at - -
          Rat Genome 230 2.0 Array 1387447_at - -
          Rat Genome 230 2.0 Array 1387447_PM_at - -
          Affymetrix HT Rat Focus Array 1387447_at - -
          Affymetrix HT Rat Focus Array 1388997_at - -
          Affymetrix HT Rat Focus Array 1388997_PM_at - -
          Affymetrix HT Rat Focus Array 1387447_PM_at - -
          Rat Expression Set 230 1388997_PM_at - -
          Rat Expression Set 230 1388997_at - -
          Rat Expression Set 230 1387447_PM_at - -
          Rat Expression Set 230 1387447_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906944 - -
          Rat Genome U34 Set L12382_at - -
          Rat Genome U34 Set rc_AI059910_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20957513 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360230 - -
          RatRef-12 Expression BeadChip ILMN_64859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline