Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02349475_m1
          See other XKR9 GE Assays ›
          Gene Symbol:
          Xkr9
          Gene Name:
          XK, Kell blood group complex subunit-related family, member 9
          Gene Aliases:
          XRG9
          Chromosome Location:
          Chr.5: 4946332 - 4975436 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02349475_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xkr9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012229.1 NP_001012229.1 3 - 4 956 122
          XM_006237733.2 XP_006237795.1 3 - 4 1039 122
          GenBank mRNA AY534263.1 - 3 - 4 956 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002153.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xkr9

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P103179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139440 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P103179 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P103179 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874966 - -
          Affymetrix Rat Gene 1.0 ST Array 10874964 - -
          Affymetrix Rat Gene 1.0 ST Array 10874965 - -
          Affymetrix Rat Gene 1.0 ST Array 10874967 - -
          Affymetrix Rat Gene 1.1 ST Array 10874963 - -
          Affymetrix Rat Clariom S Assay PSR0500016375.rn.1 TC0500002153.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10874963 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369478 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0