Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02396101_g1
          See other PHF11 GE Assays ›
          Gene Symbol:
          Phf11
          Gene Name:
          PHD finger protein 11
          Gene Aliases:
          Phf11a
          Chromosome Location:
          Chr.15: 38692811 - 39683409 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02396101_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phf11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024272.1 NP_001019443.1 6 - 7 526 107
          GenBank mRNA BC088433.1 - 6 - 7 526 107
          FQ230234.1 - 2 - 3 107
          FQ232641.1 - 2 - 3 106 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001792.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phf11

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P151851 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P243897 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P028371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P243897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P151851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P243897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P243897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P151851 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10168 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784234 - -
          Affymetrix Rat Gene 1.0 ST Array 10784236 - -
          Affymetrix Rat Gene 1.0 ST Array 10784235 - -
          Affymetrix Rat Gene 1.0 ST Array 10784232 - -
          Affymetrix Rat Gene 1.0 ST Array 10784228 - -
          Affymetrix Rat Gene 1.0 ST Array 10784237 - -
          Affymetrix Rat Gene 1.1 ST Array 10784227 - -
          Rat Genome 230 2.0 Array 1382546_PM_at - -
          Rat Genome 230 2.0 Array 1382546_at - -
          Affymetrix HT Rat Focus Array 1382546_PM_at - -
          Affymetrix HT Rat Focus Array 1382546_at - -
          Affymetrix Rat Clariom S Assay PSR1500011978.rn.1 TC1500001792.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011977.rn.1 TC1500001792.rn.1 RTA TC
          Rat Expression Set 230 1382546_PM_at - -
          Rat Expression Set 230 1382546_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784227 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360511 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0