Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02396226_m1
          See other APCDD1L GE Assays ›
          Gene Symbol:
          Apcdd1l
          Gene Name:
          adenomatosis polyposis coli down-regulated 1-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 171883156 - 171958156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02396226_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apcdd1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006235711.2 XP_006235773.1 2 - 3 683 79
          XM_008762580.1 XP_008760802.1 2 - 3 2765 79
          GenBank mRNA BC081904.1 - 523 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004960.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66727 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148396 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27389 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102547 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852166 - -
          Affymetrix Rat Gene 1.0 ST Array 10852164 - -
          Affymetrix Rat Gene 1.0 ST Array 10852161 - -
          Affymetrix Rat Gene 1.0 ST Array 10852165 - -
          Affymetrix Rat Gene 1.0 ST Array 10852167 - -
          Affymetrix Rat Clariom S Assay PSR0300037770.rn.1 TC0300004960.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10852160 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0