Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02769895_g1
          See other SULT1C2 GE Assays ›
          Gene Symbol:
          Sult1c2 RGD1562392
          Gene Name:
          sulfotransferase family, cytosolic, 1C, member 2
          similar to Sulfotransferase K1 (rSULT1C2)
          Gene Aliases:
          ST1C2
          -
          Chromosome Location:
          Chr.9: 4654278 - 4685416 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 4241300 - 4280782 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn02769895_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sult1c2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133547.4 NP_598231.3 10 1534 87
          XM_006244242.2 XP_006244304.1 9 1510 87
          XM_006244243.2 XP_006244305.1 9 1502 87
          XM_006244245.2 XP_006244307.1 9 1497 87
          XM_006244246.2 XP_006244308.1 9 1485 87
          XM_006244248.2 XP_006244310.1 10 1627 87
          XM_006244249.2 XP_006244311.1 8 1436 87
          XM_006244250.2 XP_006244312.1 8 1545 87
          XM_008766738.1 XP_008764960.1 9 1515 87
          XM_008766739.1 XP_008764961.1 9 1478 87
          XM_008766740.1 XP_008764962.1 9 1508 87
          XM_008766741.1 XP_008764963.1 9 1503 87
          XM_008766742.1 XP_008764964.1 9 1490 87
          XM_008766743.1 XP_008764965.1 9 1533 87
          XM_008766744.1 XP_008764966.1 8 1423 87
          XM_008766745.1 XP_008764967.1 8 1411 87
          GenBank mRNA BC103636.1 - 10 1534 87
          Gene Symbol: RGD1562392
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750614.3 XP_003750662.1 8 1443 87
          XM_008766791.1 XP_008765013.1 8 1422 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001406.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sult1c2
          Click here to find siRNAs for Gene RGD1562392

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          sulfation
          sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76185 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001334 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001334 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P548024 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P788758 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700596731 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97437 - -
          Agilent Rat cDNA Microarray (G4105A) 700596731 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10921241 - -
          Affymetrix Rat Gene 1.0 ST Array 10926135 - -
          Affymetrix Rat Gene 1.0 ST Array 10926099 - -
          Affymetrix Rat Gene 1.0 ST Array 10926100 - -
          Affymetrix Rat Gene 1.0 ST Array 10926102 - -
          Affymetrix Rat Gene 1.0 ST Array 10926137 - -
          Affymetrix Rat Gene 1.0 ST Array 10926126 - -
          Affymetrix Rat Gene 1.0 ST Array 10921246 - -
          Affymetrix Rat Gene 1.0 ST Array 10921242 - -
          Affymetrix Rat Gene 1.0 ST Array 10921247 - -
          Affymetrix Rat Gene 1.0 ST Array 10926101 - -
          Affymetrix Rat Gene 1.0 ST Array 10926104 - -
          Affymetrix Rat Gene 1.0 ST Array 10926133 - -
          Affymetrix Rat Gene 1.0 ST Array 10926138 - -
          Affymetrix Rat Gene 1.0 ST Array 10926103 - -
          Affymetrix Rat Gene 1.0 ST Array 10926132 - -
          Affymetrix Rat Gene 1.0 ST Array 10926130 - -
          Affymetrix Rat Gene 1.0 ST Array 10926136 - -
          Affymetrix Rat Gene 1.0 ST Array 10926129 - -
          Affymetrix Rat Gene 1.0 ST Array 10926134 - -
          Affymetrix Rat Gene 1.0 ST Array 10921248 - -
          Affymetrix Rat Gene 1.0 ST Array 10926139 - -
          Affymetrix Rat Gene 1.0 ST Array 10921245 - -
          Affymetrix Rat Gene 1.0 ST Array 10921243 - -
          Affymetrix Rat Gene 1.0 ST Array 10921244 - -
          Affymetrix Rat Gene 1.1 ST Array 10926098 - -
          Affymetrix Rat Gene 1.1 ST Array 10921239 - -
          Affymetrix Rat Gene 1.1 ST Array 10926125 - -
          Rat Genome 230 2.0 Array 1377672_at - -
          Rat Genome 230 2.0 Array 1377672_PM_at - -
          Rat Genome 230 2.0 Array 1370943_PM_at - -
          Rat Genome 230 2.0 Array 1369531_at - -
          Rat Genome 230 2.0 Array 1369531_PM_at - -
          Rat Genome 230 2.0 Array 1370943_at - -
          Affymetrix HT Rat Focus Array 1369531_PM_at - -
          Affymetrix HT Rat Focus Array 1369531_at - -
          Affymetrix Rat Clariom S Assay PSR0900011646.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011648.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011657.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011651.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011647.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011649.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011652.rn.1 TC0900001406.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011645.rn.1 TC0900001406.rn.1 RTA TC
          Rat Expression Set 230 1369531_PM_at - -
          Rat Expression Set 230 1369531_at - -
          Rat Expression Set 230 1377672_PM_at - -
          Rat Expression Set 230 1377672_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926125 - -
          Affymetrix Rat Gene 1.0 ST Array 10926098 - -
          Affymetrix Rat Gene 1.0 ST Array 10921239 - -
          Rat Genome U34 Set rc_AI007877_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363504 - -
          RatRef-12 Expression BeadChip ILMN_57863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0